Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88843729-88843996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC8
Entrez Gene ID
123016
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
BBS8, RP51
UniGene
Hs.303055
Gene Chromosome Location
Chr.14: 88824153-88877996 on Build GRCh38
Transcripts
NM_001288781.1 NM_001288782.1 NM_001288783.1 NM_144596.3 NM_198309.3 NM_198310.3 XM_006720035.1 XM_006720037.2 XM_011536432.1 XM_011536433.1 XM_011536434.1 XM_011536435.2 XM_017020977.1 XM_017020978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
958567 c.597C>A p.I199I Chr.14: 88843823-88843823 on Build GRCh38
5761612 c.594+6T>A p.? Chr.14: 88843856-88843856 on Build GRCh38
5761611 c.624+6T>A p.? Chr.14: 88843856-88843856 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754623592 Chr.14: 88843773-88843773 on Build GRCh38
rs768111024 Chr.14: 88843779-88843779 on Build GRCh38
rs758334591 Chr.14: 88843762-88843762 on Build GRCh38
rs1013412005 Chr.14: 88843948-88843948 on Build GRCh38
rs587777806 Chr.14: 88843815-88843820 on Build GRCh38
rs747642574 Chr.14: 88843791-88843792 on Build GRCh38
rs1022322949 Chr.14: 88843900-88843900 on Build GRCh38
rs748742717 Chr.14: 88843835-88843835 on Build GRCh38
rs780594621 Chr.14: 88843774-88843775 on Build GRCh38
rs777519251 Chr.14: 88843857-88843857 on Build GRCh38
rs587777808 Chr.14: 88843851-88843851 on Build GRCh38
rs757133928 Chr.14: 88843875-88843875 on Build GRCh38
rs764515706 Chr.14: 88843762-88843762 on Build GRCh38
rs897691117 Chr.14: 88843944-88843944 on Build GRCh38
rs770296360 Chr.14: 88843895-88843895 on Build GRCh38
rs766384679 Chr.14: 88843816-88843816 on Build GRCh38
rs202163736 Chr.14: 88843788-88843788 on Build GRCh38
rs1018209116 Chr.14: 88843833-88843833 on Build GRCh38
rs745806578 Chr.14: 88843882-88843882 on Build GRCh38
rs1036262794 Chr.14: 88843919-88843919 on Build GRCh38
rs182035116 Chr.14: 88843876-88843876 on Build GRCh38
rs746842052 Chr.14: 88843860-88843860 on Build GRCh38
rs201515510 Chr.14: 88843761-88843762 on Build GRCh38
rs753995884 Chr.14: 88843822-88843822 on Build GRCh38
rs200633752 Chr.14: 88843778-88843778 on Build GRCh38
rs761421321 Chr.14: 88843872-88843872 on Build GRCh38
rs756279546 Chr.14: 88843791-88843791 on Build GRCh38
rs12890993 Chr.14: 88843917-88843917 on Build GRCh38

View Full Product Details