Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 27308909-27309158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WSB1
Entrez Gene ID
26118
Gene Name
WD repeat and SOCS box containing 1
Gene Aliases
SWIP1, WSB-1
UniGene
Hs.446017
Gene Chromosome Location
Chr.17: 27294080-27313631 on Build GRCh38
Transcripts
NM_015626.8 NM_134265.2 XM_005257963.3 XM_017024433.1 XM_017024434.1 XM_017024435.1 XM_017024436.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7187763 c.770A>G p.H257R Chr.17: 27309158-27309158 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs549770437 Chr.17: 27309069-27309069 on Build GRCh38
rs370133719 Chr.17: 27309065-27309065 on Build GRCh38
rs560517627 Chr.17: 27309034-27309034 on Build GRCh38
rs896873782 Chr.17: 27308941-27308941 on Build GRCh38
rs753214233 Chr.17: 27309059-27309060 on Build GRCh38
rs181365666 Chr.17: 27308955-27308955 on Build GRCh38
rs748515416 Chr.17: 27309104-27309104 on Build GRCh38
rs777492720 Chr.17: 27308975-27308978 on Build GRCh38
rs751347915 Chr.17: 27309132-27309132 on Build GRCh38
rs769090093 Chr.17: 27309070-27309070 on Build GRCh38
rs767852737 Chr.17: 27309057-27309058 on Build GRCh38
rs762461684 Chr.17: 27309128-27309128 on Build GRCh38
rs1017880841 Chr.17: 27309045-27309045 on Build GRCh38
rs759128556 Chr.17: 27309116-27309116 on Build GRCh38
rs997890689 Chr.17: 27308965-27308965 on Build GRCh38
rs144755928 Chr.17: 27309108-27309108 on Build GRCh38
rs769809843 Chr.17: 27309061-27309061 on Build GRCh38
rs1030452013 Chr.17: 27309009-27309009 on Build GRCh38
rs536716517 Chr.17: 27309126-27309126 on Build GRCh38
rs186958628 Chr.17: 27309054-27309054 on Build GRCh38
rs764771559 Chr.17: 27309119-27309119 on Build GRCh38
rs774628034 Chr.17: 27309076-27309076 on Build GRCh38
rs146317362 Chr.17: 27309049-27309049 on Build GRCh38
rs750615999 Chr.17: 27309006-27309006 on Build GRCh38
rs555105758 Chr.17: 27309129-27309129 on Build GRCh38
rs199550651 Chr.17: 27309105-27309105 on Build GRCh38
rs775236659 Chr.17: 27309125-27309125 on Build GRCh38
rs751533375 Chr.17: 27309053-27309053 on Build GRCh38
rs986777473 Chr.17: 27309039-27309039 on Build GRCh38
rs746009536 Chr.17: 27309058-27309058 on Build GRCh38
rs766761433 Chr.17: 27309130-27309130 on Build GRCh38
rs138223081 Chr.17: 27309055-27309055 on Build GRCh38

View Full Product Details