Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 81590748-81591011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLE1
Entrez Gene ID
7088
Gene Name
transducin like enhancer of split 1
Gene Aliases
ESG, ESG1, GRG1
UniGene
Hs.197320
Gene Chromosome Location
Chr.9: 81583683-81689535 on Build GRCh38
Transcripts
NM_001303103.1 NM_001303104.1 NM_005077.4 XM_005252151.1 XM_005252152.1 XM_005252153.1 XM_005252154.1 XM_005252156.2 XM_005252162.1 XM_005252163.2 XM_006717258.1 XM_006717259.3 XM_006717260.1 XM_006717261.2 XM_006717262.1 XM_006717263.1 XM_011518951.2 XM_017015064.1 XM_017015065.1 XM_017015066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1110440 c.1707G>T p.E569D Chr.9: 81590927-81590927 on Build GRCh38
6356141 c.1817A>G p.Q606R Chr.9: 81590817-81590817 on Build GRCh38
5808367 c.1721C>T p.A574V Chr.9: 81590913-81590913 on Build GRCh38
1463182 c.1660T>G p.L554V Chr.9: 81590974-81590974 on Build GRCh38
6409577 c.1798G>A p.V600M Chr.9: 81590836-81590836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775260053 Chr.9: 81590960-81590960 on Build GRCh38
rs767262164 Chr.9: 81590883-81590883 on Build GRCh38
rs762753346 Chr.9: 81590848-81590848 on Build GRCh38
rs187303459 Chr.9: 81590772-81590772 on Build GRCh38
rs796655137 Chr.9: 81590952-81590952 on Build GRCh38
rs758543014 Chr.9: 81590927-81590927 on Build GRCh38
rs980854823 Chr.9: 81590893-81590893 on Build GRCh38
rs531274386 Chr.9: 81590788-81590788 on Build GRCh38
rs368595624 Chr.9: 81590890-81590890 on Build GRCh38
rs764412759 Chr.9: 81590900-81590900 on Build GRCh38
rs759110088 Chr.9: 81590955-81590955 on Build GRCh38
rs938335615 Chr.9: 81590915-81590915 on Build GRCh38
rs747416621 Chr.9: 81590780-81590793 on Build GRCh38
rs143199419 Chr.9: 81590921-81590921 on Build GRCh38
rs749798894 Chr.9: 81590776-81590776 on Build GRCh38
rs773182013 Chr.9: 81590840-81590840 on Build GRCh38
rs771456157 Chr.9: 81590780-81590781 on Build GRCh38
rs780219889 Chr.9: 81590931-81590931 on Build GRCh38
rs371313796 Chr.9: 81590793-81590793 on Build GRCh38
rs778808759 Chr.9: 81590778-81590778 on Build GRCh38
rs373720466 Chr.9: 81590773-81590773 on Build GRCh38
rs372013873 Chr.9: 81590909-81590909 on Build GRCh38
rs948261369 Chr.9: 81590946-81590946 on Build GRCh38
rs773935551 Chr.9: 81590852-81590852 on Build GRCh38
rs777395152 Chr.9: 81590941-81590941 on Build GRCh38
rs369331200 Chr.9: 81590942-81590942 on Build GRCh38
rs928338058 Chr.9: 81590906-81590906 on Build GRCh38
rs770937009 Chr.9: 81590849-81590849 on Build GRCh38
rs375550899 Chr.9: 81590882-81590882 on Build GRCh38
rs763838771 Chr.9: 81590986-81590986 on Build GRCh38
rs999648667 Chr.9: 81590976-81590976 on Build GRCh38
rs757664691 Chr.9: 81590912-81590912 on Build GRCh38
rs774927089 Chr.9: 81590861-81590861 on Build GRCh38
rs146691736 Chr.9: 81590940-81590940 on Build GRCh38
rs373236831 Chr.9: 81590950-81590950 on Build GRCh38
rs917099895 Chr.9: 81590877-81590877 on Build GRCh38
rs982494121 Chr.9: 81590896-81590896 on Build GRCh38
rs577470517 Chr.9: 81590812-81590812 on Build GRCh38
rs148605258 Chr.9: 81590916-81590916 on Build GRCh38
rs752460555 Chr.9: 81590885-81590885 on Build GRCh38
rs144048030 Chr.9: 81590954-81590954 on Build GRCh38
rs757335878 Chr.9: 81590777-81590777 on Build GRCh38
rs760195159 Chr.9: 81590981-81590981 on Build GRCh38
rs150975417 Chr.9: 81590957-81590957 on Build GRCh38
rs968410043 Chr.9: 81590774-81590774 on Build GRCh38
rs201024387 Chr.9: 81590836-81590836 on Build GRCh38
rs201873489 Chr.9: 81590922-81590922 on Build GRCh38
rs8782 Chr.9: 81590945-81590945 on Build GRCh38
rs762400678 Chr.9: 81590933-81590933 on Build GRCh38
rs367552658 Chr.9: 81590789-81590789 on Build GRCh38
rs769518153 Chr.9: 81590816-81590816 on Build GRCh38
rs996592604 Chr.9: 81590968-81590968 on Build GRCh38

View Full Product Details