Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95680411-95680670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCTN3
Entrez Gene ID
26123
Gene Name
tectonic family member 3
Gene Aliases
C10orf61, JBTS18, OFD4, TECT3
UniGene
Hs.438991
Gene Chromosome Location
Chr.10: 95663396-95694143 on Build GRCh38
Transcripts
NM_001143973.1 NM_015631.5 XM_005269690.1 XM_011539627.1 XM_011539628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6798956 c.985G>A p.V329I Chr.10: 95680543-95680543 on Build GRCh38
6798949 c.1556A>G p.Q519R Chr.10: 95680560-95680560 on Build GRCh38
4732868 c.946A>G p.I316V Chr.10: 95680582-95680582 on Build GRCh38
6798948 c.968A>G p.Q323R Chr.10: 95680560-95680560 on Build GRCh38
1971258 c.1020A>G p.S340S Chr.10: 95680508-95680508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746521997 Chr.10: 95680575-95680575 on Build GRCh38
rs751821516 Chr.10: 95680511-95680511 on Build GRCh38
rs139612049 Chr.10: 95680572-95680572 on Build GRCh38
rs767733835 Chr.10: 95680596-95680596 on Build GRCh38
rs371502935 Chr.10: 95680613-95680613 on Build GRCh38
rs745364410 Chr.10: 95680488-95680488 on Build GRCh38
rs779860328 Chr.10: 95680586-95680586 on Build GRCh38
rs751864135 Chr.10: 95680559-95680559 on Build GRCh38
rs759717531 Chr.10: 95680593-95680593 on Build GRCh38
rs745562513 Chr.10: 95680551-95680551 on Build GRCh38
rs1017501824 Chr.10: 95680542-95680542 on Build GRCh38
rs768137956 Chr.10: 95680487-95680487 on Build GRCh38
rs201367852 Chr.10: 95680493-95680493 on Build GRCh38
rs755119467 Chr.10: 95680450-95680450 on Build GRCh38
rs576518466 Chr.10: 95680533-95680533 on Build GRCh38
rs763929909 Chr.10: 95680633-95680633 on Build GRCh38
rs749019337 Chr.10: 95680535-95680535 on Build GRCh38
rs756071891 Chr.10: 95680468-95680468 on Build GRCh38
rs558867321 Chr.10: 95680589-95680589 on Build GRCh38
rs781327291 Chr.10: 95680465-95680480 on Build GRCh38
rs755903123 Chr.10: 95680541-95680542 on Build GRCh38
rs766507489 Chr.10: 95680504-95680504 on Build GRCh38
rs773836497 Chr.10: 95680585-95680585 on Build GRCh38
rs912033506 Chr.10: 95680631-95680631 on Build GRCh38
rs562300842 Chr.10: 95680471-95680471 on Build GRCh38
rs367614437 Chr.10: 95680446-95680446 on Build GRCh38
rs746082339 Chr.10: 95680537-95680538 on Build GRCh38
rs777395099 Chr.10: 95680485-95680485 on Build GRCh38
rs781078051 Chr.10: 95680457-95680457 on Build GRCh38
rs147156502 Chr.10: 95680626-95680626 on Build GRCh38
rs1030704966 Chr.10: 95680517-95680517 on Build GRCh38
rs765803620 Chr.10: 95680500-95680500 on Build GRCh38
rs749117268 Chr.10: 95680486-95680486 on Build GRCh38
rs757139578 Chr.10: 95680544-95680544 on Build GRCh38
rs536802100 Chr.10: 95680462-95680462 on Build GRCh38
rs1026823695 Chr.10: 95680523-95680523 on Build GRCh38
rs748135693 Chr.10: 95680464-95680464 on Build GRCh38
rs149728983 Chr.10: 95680588-95680588 on Build GRCh38
rs989305259 Chr.10: 95680578-95680578 on Build GRCh38
rs756561177 Chr.10: 95680524-95680524 on Build GRCh38
rs752719561 Chr.10: 95680597-95680597 on Build GRCh38
rs774722961 Chr.10: 95680592-95680592 on Build GRCh38
rs755942179 Chr.10: 95680525-95680525 on Build GRCh38
rs760863463 Chr.10: 95680606-95680606 on Build GRCh38
rs150579201 Chr.10: 95680495-95680495 on Build GRCh38
rs755029867 Chr.10: 95680521-95680521 on Build GRCh38
rs12784530 Chr.10: 95680637-95680637 on Build GRCh38
rs767539312 Chr.10: 95680522-95680522 on Build GRCh38
rs763433183 Chr.10: 95680501-95680501 on Build GRCh38
rs771745084 Chr.10: 95680490-95680490 on Build GRCh38
rs376520003 Chr.10: 95680549-95680549 on Build GRCh38
rs770188453 Chr.10: 95680582-95680582 on Build GRCh38

View Full Product Details