Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24299793-24300066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM28
Entrez Gene ID
10863
Gene Name
ADAM metallopeptidase domain 28
Gene Aliases
ADAM 28, MDC-L, MDCL, eMDC II, eMDCII
UniGene
Hs.174030
Gene Chromosome Location
Chr.8: 24294040-24359018 on Build GRCh38
Transcripts
NM_001304351.1 NM_014265.5 NM_021777.4 NR_130709.1 NR_130710.1 XM_005273380.3 XM_005273382.1 XM_006716273.3 XM_006716274.1 XM_011544367.2 XM_011544368.2 XM_011544369.2 XM_011544370.2 XM_017012974.1 XM_017012975.1 XM_017012976.1 NR_125808.1

Gene Symbol
LOC101929294
Entrez Gene ID
101929294
Gene Name
uncharacterized LOC101929294
Gene Aliases
-
UniGene
Hs.637673
Gene Chromosome Location
Chr.8: 24295814-24548618 on Build GRCh38
Transcripts
NM_001304351.1 NM_014265.5 NM_021777.4 NR_130709.1 NR_130710.1 XM_005273380.3 XM_005273382.1 XM_006716273.3 XM_006716274.1 XM_011544367.2 XM_011544368.2 XM_011544369.2 XM_011544370.2 XM_017012974.1 XM_017012975.1 XM_017012976.1 NR_125808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866015 c.56T>A p.I19K Chr.8: 24299983-24299983 on Build GRCh38
6866016 c.56T>A p.I19K Chr.8: 24299983-24299983 on Build GRCh38
259316 c.104G>T p.R35I Chr.8: 24300031-24300031 on Build GRCh38
1699828 c.61G>A p.E21K Chr.8: 24299988-24299988 on Build GRCh38
4909525 c.119A>G p.H40R Chr.8: 24300046-24300046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375587519 Chr.8: 24300032-24300032 on Build GRCh38
rs769981343 Chr.8: 24300029-24300029 on Build GRCh38
rs767817464 Chr.8: 24299941-24299941 on Build GRCh38
rs754885982 Chr.8: 24299968-24299969 on Build GRCh38
rs374072282 Chr.8: 24299938-24299938 on Build GRCh38
rs143371817 Chr.8: 24299988-24299988 on Build GRCh38
rs961043715 Chr.8: 24299846-24299846 on Build GRCh38
rs918241212 Chr.8: 24299888-24299888 on Build GRCh38
rs750451322 Chr.8: 24299962-24299963 on Build GRCh38
rs549811083 Chr.8: 24299949-24299949 on Build GRCh38
rs776606602 Chr.8: 24299820-24299820 on Build GRCh38
rs112530601 Chr.8: 24299965-24299965 on Build GRCh38
rs867069414 Chr.8: 24300012-24300012 on Build GRCh38
rs756940015 Chr.8: 24299951-24299951 on Build GRCh38
rs112820180 Chr.8: 24300010-24300010 on Build GRCh38
rs369332654 Chr.8: 24300041-24300041 on Build GRCh38
rs963656371 Chr.8: 24299860-24299860 on Build GRCh38
rs781283001 Chr.8: 24299950-24299954 on Build GRCh38
rs781182940 Chr.8: 24299997-24299997 on Build GRCh38
rs750069237 Chr.8: 24300016-24300016 on Build GRCh38
rs762958280 Chr.8: 24300040-24300040 on Build GRCh38
rs911600151 Chr.8: 24299818-24299818 on Build GRCh38
rs1044158829 Chr.8: 24299917-24299917 on Build GRCh38
rs201353852 Chr.8: 24299929-24299929 on Build GRCh38
rs766858101 Chr.8: 24299934-24299934 on Build GRCh38
rs749959262 Chr.8: 24299962-24299962 on Build GRCh38
rs778619154 Chr.8: 24299956-24299956 on Build GRCh38
rs564835727 Chr.8: 24299891-24299891 on Build GRCh38
rs757867509 Chr.8: 24299969-24299969 on Build GRCh38
rs747921683 Chr.8: 24299999-24299999 on Build GRCh38
rs761265746 Chr.8: 24299885-24299885 on Build GRCh38
rs779858610 Chr.8: 24299980-24299980 on Build GRCh38
rs922064427 Chr.8: 24299890-24299890 on Build GRCh38
rs1016895993 Chr.8: 24299822-24299822 on Build GRCh38
rs773474621 Chr.8: 24300033-24300033 on Build GRCh38
rs375265578 Chr.8: 24299977-24299977 on Build GRCh38
rs764760622 Chr.8: 24299982-24299982 on Build GRCh38

View Full Product Details