Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202467504-202467731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR2
Entrez Gene ID
659
Gene Name
bone morphogenetic protein receptor type 2
Gene Aliases
BMPR-II, BMPR3, BMR2, BRK-3, POVD1, PPH1, T-ALK
UniGene
Hs.471119
Gene Chromosome Location
Chr.2: 202376310-202567751 on Build GRCh38
Transcripts
NM_001204.6 XM_011511687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612059 c.334A>T p.T112S Chr.2: 202467605-202467605 on Build GRCh38
6612058 c.347G>T p.C116F Chr.2: 202467618-202467618 on Build GRCh38
7357608 c.329A>G p.N110S Chr.2: 202467600-202467600 on Build GRCh38
6503979 c.404A>G p.D135G Chr.2: 202467675-202467675 on Build GRCh38
4544693 c.361G>A p.D121N Chr.2: 202467632-202467632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781346279 Chr.2: 202467542-202467542 on Build GRCh38
rs761096267 Chr.2: 202467588-202467588 on Build GRCh38
rs776904933 Chr.2: 202467594-202467594 on Build GRCh38
rs900978149 Chr.2: 202467680-202467680 on Build GRCh38
rs866080270 Chr.2: 202467683-202467683 on Build GRCh38
rs768903998 Chr.2: 202467564-202467564 on Build GRCh38
rs749485755 Chr.2: 202467559-202467559 on Build GRCh38
rs750321249 Chr.2: 202467575-202467577 on Build GRCh38
rs137852743 Chr.2: 202467625-202467625 on Build GRCh38
rs750326466 Chr.2: 202467688-202467688 on Build GRCh38
rs863223425 Chr.2: 202467566-202467566 on Build GRCh38
rs766729194 Chr.2: 202467593-202467593 on Build GRCh38
rs371271567 Chr.2: 202467685-202467685 on Build GRCh38
rs779736124 Chr.2: 202467557-202467557 on Build GRCh38
rs369156981 Chr.2: 202467682-202467682 on Build GRCh38
rs751111275 Chr.2: 202467679-202467679 on Build GRCh38
rs773638601 Chr.2: 202467584-202467584 on Build GRCh38
rs772560246 Chr.2: 202467581-202467581 on Build GRCh38
rs774390535 Chr.2: 202467573-202467573 on Build GRCh38
rs942477672 Chr.2: 202467543-202467543 on Build GRCh38
rs372058084 Chr.2: 202467579-202467579 on Build GRCh38
rs137852750 Chr.2: 202467638-202467638 on Build GRCh38
rs986134798 Chr.2: 202467598-202467598 on Build GRCh38
rs375174261 Chr.2: 202467673-202467673 on Build GRCh38
rs140683387 Chr.2: 202467547-202467547 on Build GRCh38
rs756036199 Chr.2: 202467700-202467700 on Build GRCh38
rs745800266 Chr.2: 202467546-202467546 on Build GRCh38
rs863223426 Chr.2: 202467648-202467648 on Build GRCh38
rs762646803 Chr.2: 202467628-202467628 on Build GRCh38

View Full Product Details