Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35499968-35500193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TULP1
Entrez Gene ID
7287
Gene Name
tubby like protein 1
Gene Aliases
LCA15, RP14, TUBL1
UniGene
Hs.485208
Gene Chromosome Location
Chr.6: 35497874-35512902 on Build GRCh38
Transcripts
NM_001289395.1 NM_003322.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3626012 c.1370G>A p.S457N Chr.6: 35500106-35500106 on Build GRCh38
1329728 c.1470C>A p.N490K Chr.6: 35500006-35500006 on Build GRCh38
3626011 c.1408G>A p.D470N Chr.6: 35500068-35500068 on Build GRCh38
5157176 c.1346G>A p.R449H Chr.6: 35500130-35500130 on Build GRCh38
3702986 c.1418G>C p.G473A Chr.6: 35500058-35500058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753328447 Chr.6: 35500161-35500161 on Build GRCh38
rs121909074 Chr.6: 35500005-35500005 on Build GRCh38
rs533734109 Chr.6: 35499991-35499991 on Build GRCh38
rs943902351 Chr.6: 35500080-35500080 on Build GRCh38
rs121909077 Chr.6: 35500032-35500032 on Build GRCh38
rs746118991 Chr.6: 35500039-35500039 on Build GRCh38
rs760326438 Chr.6: 35500092-35500092 on Build GRCh38
rs138200747 Chr.6: 35500115-35500115 on Build GRCh38
rs775196326 Chr.6: 35500087-35500087 on Build GRCh38
rs138400030 Chr.6: 35500146-35500146 on Build GRCh38
rs985811337 Chr.6: 35500045-35500045 on Build GRCh38
rs982387647 Chr.6: 35500019-35500019 on Build GRCh38
rs181172036 Chr.6: 35500143-35500143 on Build GRCh38
rs772500923 Chr.6: 35500149-35500149 on Build GRCh38
rs368559087 Chr.6: 35500036-35500036 on Build GRCh38
rs370691307 Chr.6: 35500111-35500111 on Build GRCh38
rs953685924 Chr.6: 35500025-35500025 on Build GRCh38
rs546453354 Chr.6: 35500118-35500118 on Build GRCh38
rs141666148 Chr.6: 35500132-35500132 on Build GRCh38
rs777719248 Chr.6: 35500117-35500117 on Build GRCh38
rs754507488 Chr.6: 35499997-35499997 on Build GRCh38
rs753517944 Chr.6: 35500098-35500098 on Build GRCh38
rs62636511 Chr.6: 35500010-35500010 on Build GRCh38
rs762348395 Chr.6: 35500077-35500077 on Build GRCh38
rs766088997 Chr.6: 35500095-35500095 on Build GRCh38
rs772035344 Chr.6: 35500018-35500018 on Build GRCh38
rs749502078 Chr.6: 35500142-35500142 on Build GRCh38
rs141980901 Chr.6: 35499990-35499990 on Build GRCh38
rs1036874387 Chr.6: 35500082-35500082 on Build GRCh38
rs61734562 Chr.6: 35500135-35500135 on Build GRCh38
rs773507942 Chr.6: 35500091-35500091 on Build GRCh38
rs771867194 Chr.6: 35500086-35500086 on Build GRCh38
rs749392203 Chr.6: 35500049-35500049 on Build GRCh38
rs868745568 Chr.6: 35500102-35500102 on Build GRCh38
rs281865533 Chr.6: 35500101-35500101 on Build GRCh38
rs774767185 Chr.6: 35500070-35500070 on Build GRCh38
rs773706008 Chr.6: 35500160-35500160 on Build GRCh38
rs146311742 Chr.6: 35500031-35500031 on Build GRCh38
rs138537513 Chr.6: 35500043-35500043 on Build GRCh38
rs866341403 Chr.6: 35500068-35500068 on Build GRCh38
rs41270076 Chr.6: 35500114-35500114 on Build GRCh38
rs770079177 Chr.6: 35500042-35500042 on Build GRCh38
rs112710652 Chr.6: 35500083-35500083 on Build GRCh38
rs766709626 Chr.6: 35500165-35500165 on Build GRCh38
rs767899721 Chr.6: 35500090-35500090 on Build GRCh38
rs774448249 Chr.6: 35500144-35500144 on Build GRCh38
rs121909075 Chr.6: 35500100-35500100 on Build GRCh38
rs142578700 Chr.6: 35500069-35500069 on Build GRCh38

View Full Product Details