Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41439351-41439612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5SL
Entrez Gene ID
55101
Gene Name
ATP5S like
Gene Aliases
-
UniGene
Hs.351099
Gene Chromosome Location
Chr.19: 41431318-41447822 on Build GRCh38
Transcripts
NM_001167867.1 NM_001167868.1 NM_001167869.1 NM_001167870.1 NM_001167871.1 NM_001320838.1 NM_001320839.1 NM_001320840.1 NM_001320841.1 NM_001320844.1 NM_018035.2 NR_030765.1 NR_135476.1 NR_135477.1 NR_135478.1 XM_011527065.2

SNP Detail

SNP ID Chromosome Location
rs782567554 Chr.19: 41439572-41439572 on Build GRCh38
rs932951798 Chr.19: 41439497-41439497 on Build GRCh38
rs201128960 Chr.19: 41439476-41439476 on Build GRCh38
rs782104953 Chr.19: 41439507-41439507 on Build GRCh38
rs888823082 Chr.19: 41439559-41439559 on Build GRCh38
rs187336429 Chr.19: 41439494-41439494 on Build GRCh38
rs970172981 Chr.19: 41439423-41439423 on Build GRCh38
rs150658847 Chr.19: 41439590-41439590 on Build GRCh38
rs56058717 Chr.19: 41439399-41439399 on Build GRCh38
rs536331141 Chr.19: 41439502-41439502 on Build GRCh38
rs571928438 Chr.19: 41439410-41439410 on Build GRCh38
rs576177602 Chr.19: 41439573-41439573 on Build GRCh38
rs782070677 Chr.19: 41439473-41439473 on Build GRCh38
rs374426673 Chr.19: 41439580-41439580 on Build GRCh38
rs555010735 Chr.19: 41439545-41439545 on Build GRCh38
rs1051457870 Chr.19: 41439537-41439537 on Build GRCh38
rs782643700 Chr.19: 41439436-41439436 on Build GRCh38
rs915959284 Chr.19: 41439416-41439416 on Build GRCh38
rs183713873 Chr.19: 41439387-41439387 on Build GRCh38
rs370641942 Chr.19: 41439428-41439428 on Build GRCh38
rs925139386 Chr.19: 41439430-41439430 on Build GRCh38
rs781880023 Chr.19: 41439552-41439552 on Build GRCh38
rs960060172 Chr.19: 41439379-41439379 on Build GRCh38
rs548271917 Chr.19: 41439447-41439447 on Build GRCh38
rs536386175 Chr.19: 41439431-41439431 on Build GRCh38

View Full Product Details