Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107881068-107881329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC63
Entrez Gene ID
11231
Gene Name
SEC63 homolog, protein translocation regulator
Gene Aliases
DNAJC23, ERdj2, PRO2507, SEC63L
UniGene
Hs.26904
Gene Chromosome Location
Chr.6: 107867756-107958278 on Build GRCh38
Transcripts
NM_007214.4 XM_011535399.2 XM_017010218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6029926 c.1864C>T p.R622* Chr.6: 107881220-107881220 on Build GRCh38
1225097 c.1899A>C p.K633N Chr.6: 107881185-107881185 on Build GRCh38
4839206 c.1887A>C p.E629D Chr.6: 107881197-107881197 on Build GRCh38
6774326 c.1834-10delT p.? Chr.6: 107881260-107881260 on Build GRCh38
4797320 c.1836G>A p.E612E Chr.6: 107881248-107881248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768615942 Chr.6: 107881189-107881189 on Build GRCh38
rs753416822 Chr.6: 107881097-107881098 on Build GRCh38
rs201530309 Chr.6: 107881256-107881256 on Build GRCh38
rs146150463 Chr.6: 107881203-107881203 on Build GRCh38
rs762421000 Chr.6: 107881233-107881233 on Build GRCh38
rs764787923 Chr.6: 107881235-107881235 on Build GRCh38
rs138273253 Chr.6: 107881175-107881175 on Build GRCh38
rs778566521 Chr.6: 107881140-107881140 on Build GRCh38
rs925837462 Chr.6: 107881144-107881144 on Build GRCh38
rs754834382 Chr.6: 107881276-107881276 on Build GRCh38
rs774043193 Chr.6: 107881259-107881260 on Build GRCh38
rs577008691 Chr.6: 107881195-107881195 on Build GRCh38
rs761436360 Chr.6: 107881278-107881278 on Build GRCh38
rs1031651804 Chr.6: 107881095-107881095 on Build GRCh38
rs754326276 Chr.6: 107881230-107881230 on Build GRCh38
rs749930698 Chr.6: 107881240-107881240 on Build GRCh38
rs373091152 Chr.6: 107881217-107881217 on Build GRCh38
rs1044520374 Chr.6: 107881128-107881128 on Build GRCh38
rs553877910 Chr.6: 107881102-107881102 on Build GRCh38
rs750031954 Chr.6: 107881152-107881152 on Build GRCh38
rs149629810 Chr.6: 107881202-107881202 on Build GRCh38
rs779013274 Chr.6: 107881297-107881297 on Build GRCh38
rs935826770 Chr.6: 107881151-107881151 on Build GRCh38
rs745999999 Chr.6: 107881289-107881289 on Build GRCh38
rs775895484 Chr.6: 107881219-107881219 on Build GRCh38
rs759756969 Chr.6: 107881215-107881215 on Build GRCh38
rs764740802 Chr.6: 107881121-107881121 on Build GRCh38
rs768329883 Chr.6: 107881127-107881128 on Build GRCh38
rs756606612 Chr.6: 107881109-107881109 on Build GRCh38
rs746818065 Chr.6: 107881178-107881178 on Build GRCh38
rs1053005798 Chr.6: 107881184-107881184 on Build GRCh38
rs369822587 Chr.6: 107881248-107881248 on Build GRCh38
rs781164174 Chr.6: 107881191-107881191 on Build GRCh38
rs758006300 Chr.6: 107881154-107881154 on Build GRCh38
rs753027223 Chr.6: 107881108-107881108 on Build GRCh38
rs199595843 Chr.6: 107881222-107881222 on Build GRCh38
rs756890057 Chr.6: 107881136-107881136 on Build GRCh38
rs371325583 Chr.6: 107881101-107881101 on Build GRCh38
rs760998122 Chr.6: 107881228-107881228 on Build GRCh38
rs753379702 Chr.6: 107881127-107881127 on Build GRCh38
rs1011631291 Chr.6: 107881119-107881119 on Build GRCh38
rs375551086 Chr.6: 107881229-107881229 on Build GRCh38
rs751355513 Chr.6: 107881266-107881266 on Build GRCh38
rs748021398 Chr.6: 107881282-107881282 on Build GRCh38

View Full Product Details