Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119239717-119239976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP53
Entrez Gene ID
54532
Gene Name
ubiquitin specific peptidase 53
Gene Aliases
-
UniGene
Hs.431081
Gene Chromosome Location
Chr.4: 119212583-119295518 on Build GRCh38
Transcripts
NM_019050.2 XM_005263073.3 XM_005263075.2 XM_005263076.2 XM_005263077.3 XM_006714240.1 XM_011532037.2 XM_011532038.2 XM_017008312.1 XM_017008313.1 XM_017008314.1 XM_017008315.1 XM_017008316.1 XM_017008317.1 XM_017008318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3767669 c.134G>A p.S45N Chr.4: 119239893-119239893 on Build GRCh38
4607849 c.57G>T p.Q19H Chr.4: 119239816-119239816 on Build GRCh38
5995382 c.79G>A p.A27T Chr.4: 119239838-119239838 on Build GRCh38
4121617 c.135C>T p.S45S Chr.4: 119239894-119239894 on Build GRCh38
3007997 c.20T>C p.L7S Chr.4: 119239779-119239779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759401193 Chr.4: 119239787-119239787 on Build GRCh38
rs962519462 Chr.4: 119239941-119239941 on Build GRCh38
rs762005503 Chr.4: 119239816-119239816 on Build GRCh38
rs201043574 Chr.4: 119239929-119239930 on Build GRCh38
rs772051484 Chr.4: 119239791-119239791 on Build GRCh38
rs766687297 Chr.4: 119239939-119239939 on Build GRCh38
rs780407501 Chr.4: 119239831-119239831 on Build GRCh38
rs891618426 Chr.4: 119239807-119239807 on Build GRCh38
rs775459006 Chr.4: 119239895-119239895 on Build GRCh38
rs776957005 Chr.4: 119239904-119239904 on Build GRCh38
rs758746465 Chr.4: 119239829-119239829 on Build GRCh38
rs199820807 Chr.4: 119239894-119239894 on Build GRCh38
rs375688789 Chr.4: 119239798-119239798 on Build GRCh38
rs769286768 Chr.4: 119239754-119239754 on Build GRCh38
rs529585914 Chr.4: 119239927-119239927 on Build GRCh38
rs371589774 Chr.4: 119239870-119239870 on Build GRCh38
rs750992151 Chr.4: 119239818-119239818 on Build GRCh38
rs775798277 Chr.4: 119239796-119239796 on Build GRCh38
rs774479208 Chr.4: 119239782-119239782 on Build GRCh38
rs764195676 Chr.4: 119239803-119239803 on Build GRCh38
rs755751762 Chr.4: 119239855-119239855 on Build GRCh38
rs369794475 Chr.4: 119239867-119239867 on Build GRCh38
rs900628798 Chr.4: 119239852-119239852 on Build GRCh38
rs765356203 Chr.4: 119239919-119239919 on Build GRCh38
rs372405059 Chr.4: 119239771-119239771 on Build GRCh38
rs777150385 Chr.4: 119239766-119239766 on Build GRCh38
rs747853051 Chr.4: 119239752-119239752 on Build GRCh38
rs752277928 Chr.4: 119239835-119239835 on Build GRCh38
rs771528477 Chr.4: 119239874-119239874 on Build GRCh38
rs548222230 Chr.4: 119239838-119239838 on Build GRCh38
rs745815552 Chr.4: 119239876-119239876 on Build GRCh38
rs770795452 Chr.4: 119239781-119239781 on Build GRCh38
rs761791049 Chr.4: 119239912-119239912 on Build GRCh38
rs374987384 Chr.4: 119239901-119239901 on Build GRCh38
rs750941329 Chr.4: 119239930-119239930 on Build GRCh38
rs753889712 Chr.4: 119239808-119239808 on Build GRCh38
rs543833629 Chr.4: 119239765-119239765 on Build GRCh38
rs763452634 Chr.4: 119239930-119239930 on Build GRCh38
rs765698235 Chr.4: 119239817-119239817 on Build GRCh38
rs137953832 Chr.4: 119239940-119239940 on Build GRCh38
rs755419466 Chr.4: 119239943-119239943 on Build GRCh38
rs770540742 Chr.4: 119239873-119239873 on Build GRCh38

View Full Product Details