Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69090738-69090993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANTXR1
Entrez Gene ID
84168
Gene Name
anthrax toxin receptor 1
Gene Aliases
ATR, GAPO, TEM8
UniGene
Hs.165859
Gene Chromosome Location
Chr.2: 69013144-69249327 on Build GRCh38
Transcripts
NM_018153.3 NM_032208.2 NM_053034.2 XM_017005075.1 XM_017005076.1 XM_017005077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6158658 c.702A>G p.G234G Chr.2: 69090918-69090918 on Build GRCh38
1409112 c.664G>A p.E222K Chr.2: 69090880-69090880 on Build GRCh38
3055954 c.664G>A p.E222K Chr.2: 69090880-69090880 on Build GRCh38
1409113 c.664G>A p.E222K Chr.2: 69090880-69090880 on Build GRCh38
6158656 c.702A>G p.G234G Chr.2: 69090918-69090918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768766349 Chr.2: 69090879-69090879 on Build GRCh38
rs563062827 Chr.2: 69090806-69090806 on Build GRCh38
rs187291277 Chr.2: 69090957-69090957 on Build GRCh38
rs777027296 Chr.2: 69090953-69090953 on Build GRCh38
rs374173601 Chr.2: 69090958-69090958 on Build GRCh38
rs771470076 Chr.2: 69090925-69090925 on Build GRCh38
rs766595576 Chr.2: 69090809-69090809 on Build GRCh38
rs763104215 Chr.2: 69090961-69090961 on Build GRCh38
rs760203759 Chr.2: 69090945-69090945 on Build GRCh38
rs771348149 Chr.2: 69090952-69090952 on Build GRCh38
rs747729488 Chr.2: 69090907-69090907 on Build GRCh38
rs376359209 Chr.2: 69090876-69090876 on Build GRCh38
rs754373790 Chr.2: 69090823-69090823 on Build GRCh38
rs993089280 Chr.2: 69090836-69090836 on Build GRCh38
rs538935179 Chr.2: 69090797-69090797 on Build GRCh38
rs755564837 Chr.2: 69090838-69090839 on Build GRCh38
rs779957944 Chr.2: 69090851-69090851 on Build GRCh38
rs4854546 Chr.2: 69090793-69090793 on Build GRCh38
rs765708597 Chr.2: 69090827-69090827 on Build GRCh38
rs777270614 Chr.2: 69090957-69090959 on Build GRCh38
rs761159207 Chr.2: 69090808-69090808 on Build GRCh38
rs778573594 Chr.2: 69090894-69090894 on Build GRCh38
rs370320129 Chr.2: 69090826-69090826 on Build GRCh38
rs778112115 Chr.2: 69090847-69090847 on Build GRCh38
rs758401079 Chr.2: 69090838-69090838 on Build GRCh38
rs920720283 Chr.2: 69090883-69090883 on Build GRCh38
rs556643370 Chr.2: 69090848-69090848 on Build GRCh38
rs957294547 Chr.2: 69090785-69090785 on Build GRCh38
rs930622091 Chr.2: 69090897-69090897 on Build GRCh38
rs558861166 Chr.2: 69090955-69090955 on Build GRCh38
rs200029323 Chr.2: 69090956-69090956 on Build GRCh38
rs752813261 Chr.2: 69090833-69090833 on Build GRCh38
rs1031319555 Chr.2: 69090774-69090774 on Build GRCh38
rs772932392 Chr.2: 69090942-69090942 on Build GRCh38

View Full Product Details