Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 227766900-227767167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP47
Entrez Gene ID
116841
Gene Name
synaptosome associated protein 47
Gene Aliases
C1orf142, ESFI5812, HEL-S-290, HEL170, SNAP-47, SVAP1
UniGene
Hs.325081
Gene Chromosome Location
Chr.1: 227728168-227781231 on Build GRCh38
Transcripts
NM_001323930.1 NM_001323931.1 NM_001323932.1 NM_001323933.1 NM_001323934.1 NM_001323935.1 NM_053052.3 NR_136654.1 NR_136655.1 NR_136656.1 NR_136657.1 XM_006711734.3 XM_017000231.1 XM_017000232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5914783 c.1124-7C>T p.? Chr.1: 227766952-227766952 on Build GRCh38
4028826 c.1142G>A p.R381H Chr.1: 227766977-227766977 on Build GRCh38
3750931 c.1141C>T p.R381C Chr.1: 227766976-227766976 on Build GRCh38
5774151 c.1140C>T p.G380G Chr.1: 227766975-227766975 on Build GRCh38
7221354 c.1153C>T p.R385C Chr.1: 227766988-227766988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370860074 Chr.1: 227766965-227766965 on Build GRCh38
rs765555970 Chr.1: 227767042-227767042 on Build GRCh38
rs372793706 Chr.1: 227766922-227766922 on Build GRCh38
rs575623825 Chr.1: 227766932-227766932 on Build GRCh38
rs773693918 Chr.1: 227766961-227766961 on Build GRCh38
rs377682744 Chr.1: 227767023-227767023 on Build GRCh38
rs920195886 Chr.1: 227766925-227766925 on Build GRCh38
rs779821356 Chr.1: 227767093-227767093 on Build GRCh38
rs750880697 Chr.1: 227767029-227767029 on Build GRCh38
rs779512811 Chr.1: 227767026-227767026 on Build GRCh38
rs561311451 Chr.1: 227767063-227767063 on Build GRCh38
rs149591514 Chr.1: 227767015-227767015 on Build GRCh38
rs549839810 Chr.1: 227767069-227767069 on Build GRCh38
rs775831874 Chr.1: 227767135-227767135 on Build GRCh38
rs745610185 Chr.1: 227767016-227767016 on Build GRCh38
rs765313872 Chr.1: 227766980-227766980 on Build GRCh38
rs771486661 Chr.1: 227767114-227767114 on Build GRCh38
rs944050572 Chr.1: 227766966-227766966 on Build GRCh38
rs758541678 Chr.1: 227766995-227766995 on Build GRCh38
rs968658521 Chr.1: 227767067-227767067 on Build GRCh38
rs752861248 Chr.1: 227767044-227767044 on Build GRCh38
rs567491167 Chr.1: 227767133-227767133 on Build GRCh38
rs34209229 Chr.1: 227767041-227767041 on Build GRCh38
rs748906250 Chr.1: 227767094-227767094 on Build GRCh38
rs755847561 Chr.1: 227767088-227767088 on Build GRCh38
rs547268358 Chr.1: 227767049-227767049 on Build GRCh38
rs371860274 Chr.1: 227767134-227767134 on Build GRCh38
rs985435516 Chr.1: 227766959-227766959 on Build GRCh38
rs571484436 Chr.1: 227767084-227767084 on Build GRCh38
rs200001973 Chr.1: 227766999-227766999 on Build GRCh38
rs757167709 Chr.1: 227767007-227767007 on Build GRCh38
rs762302225 Chr.1: 227766942-227766942 on Build GRCh38
rs776464147 Chr.1: 227766928-227766928 on Build GRCh38
rs78405690 Chr.1: 227766988-227766988 on Build GRCh38
rs778413284 Chr.1: 227767104-227767104 on Build GRCh38
rs760505853 Chr.1: 227767025-227767025 on Build GRCh38
rs771375297 Chr.1: 227767037-227767037 on Build GRCh38
rs113250793 Chr.1: 227767095-227767095 on Build GRCh38
rs747594192 Chr.1: 227767107-227767107 on Build GRCh38
rs531705802 Chr.1: 227767068-227767068 on Build GRCh38
rs367590602 Chr.1: 227767115-227767115 on Build GRCh38
rs369206243 Chr.1: 227766921-227766921 on Build GRCh38
rs1051483082 Chr.1: 227767144-227767144 on Build GRCh38
rs1005151412 Chr.1: 227766949-227766949 on Build GRCh38
rs377565711 Chr.1: 227766939-227766939 on Build GRCh38
rs780925396 Chr.1: 227767073-227767073 on Build GRCh38
rs17851681 Chr.1: 227766976-227766976 on Build GRCh38
rs201979244 Chr.1: 227767000-227767000 on Build GRCh38
rs773745138 Chr.1: 227767030-227767030 on Build GRCh38
rs755264056 Chr.1: 227766977-227766977 on Build GRCh38
rs372397148 Chr.1: 227766920-227766920 on Build GRCh38
rs768024306 Chr.1: 227766958-227766958 on Build GRCh38
rs182307106 Chr.1: 227766967-227766967 on Build GRCh38
rs371025106 Chr.1: 227767027-227767027 on Build GRCh38
rs34538544 Chr.1: 227766989-227766989 on Build GRCh38
rs761144416 Chr.1: 227767031-227767031 on Build GRCh38
rs763167039 Chr.1: 227767048-227767048 on Build GRCh38
rs779858848 Chr.1: 227767051-227767054 on Build GRCh38
rs750244846 Chr.1: 227767085-227767085 on Build GRCh38
rs780631722 Chr.1: 227766993-227766993 on Build GRCh38
rs778864535 Chr.1: 227766927-227766927 on Build GRCh38

View Full Product Details