Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 8422403-8422676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35B3
Entrez Gene ID
51000
Gene Name
solute carrier family 35 member B3
Gene Aliases
C6orf196, CGI-19, PAPST2
UniGene
Hs.285847
Gene Chromosome Location
Chr.6: 8413066-8435606 on Build GRCh38
Transcripts
NM_001142541.2 NM_015948.4 NR_109913.1 NR_109914.1 XM_005249156.3 XM_005249157.2 XM_005249158.1 XM_006715102.3 XM_006715103.3 XM_011514648.1 XM_011514650.2 XM_017010910.1 XM_017010911.1 XM_017010912.1 XM_017010913.1 XM_017010914.1 XM_017010915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6436794 c.427G>T p.G143* Chr.6: 8422617-8422617 on Build GRCh38
3176658 c.534C>T p.C178C Chr.6: 8422510-8422510 on Build GRCh38
3411314 c.421A>G p.I141V Chr.6: 8422623-8422623 on Build GRCh38
484399 c.474G>C p.G158G Chr.6: 8422570-8422570 on Build GRCh38
3394330 c.433A>G p.T145A Chr.6: 8422611-8422611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529596595 Chr.6: 8422434-8422434 on Build GRCh38
rs754991791 Chr.6: 8422582-8422583 on Build GRCh38
rs760597579 Chr.6: 8422539-8422539 on Build GRCh38
rs373292611 Chr.6: 8422646-8422646 on Build GRCh38
rs987342097 Chr.6: 8422496-8422496 on Build GRCh38
rs931539710 Chr.6: 8422635-8422635 on Build GRCh38
rs199981680 Chr.6: 8422436-8422436 on Build GRCh38
rs907580615 Chr.6: 8422511-8422511 on Build GRCh38
rs758699761 Chr.6: 8422463-8422463 on Build GRCh38
rs766248146 Chr.6: 8422555-8422555 on Build GRCh38
rs771537703 Chr.6: 8422590-8422590 on Build GRCh38
rs748167381 Chr.6: 8422526-8422526 on Build GRCh38
rs765707833 Chr.6: 8422626-8422626 on Build GRCh38
rs370114471 Chr.6: 8422555-8422555 on Build GRCh38
rs777796568 Chr.6: 8422550-8422550 on Build GRCh38
rs777348999 Chr.6: 8422561-8422561 on Build GRCh38
rs767268440 Chr.6: 8422477-8422478 on Build GRCh38
rs755227243 Chr.6: 8422648-8422648 on Build GRCh38
rs753648540 Chr.6: 8422563-8422574 on Build GRCh38
rs758391043 Chr.6: 8422634-8422635 on Build GRCh38
rs774976524 Chr.6: 8422601-8422601 on Build GRCh38
rs761161689 Chr.6: 8422455-8422456 on Build GRCh38
rs752884499 Chr.6: 8422619-8422619 on Build GRCh38
rs769861644 Chr.6: 8422539-8422539 on Build GRCh38
rs147100909 Chr.6: 8422461-8422461 on Build GRCh38
rs770722268 Chr.6: 8422584-8422584 on Build GRCh38
rs372374762 Chr.6: 8422439-8422439 on Build GRCh38
rs773453856 Chr.6: 8422445-8422445 on Build GRCh38
rs759203429 Chr.6: 8422588-8422588 on Build GRCh38
rs138267484 Chr.6: 8422491-8422491 on Build GRCh38
rs772728080 Chr.6: 8422500-8422501 on Build GRCh38
rs996300470 Chr.6: 8422500-8422500 on Build GRCh38
rs773917543 Chr.6: 8422585-8422585 on Build GRCh38
rs780159985 Chr.6: 8422488-8422488 on Build GRCh38
rs757818556 Chr.6: 8422506-8422506 on Build GRCh38
rs954770019 Chr.6: 8422574-8422574 on Build GRCh38
rs760110146 Chr.6: 8422604-8422604 on Build GRCh38
rs973933627 Chr.6: 8422642-8422642 on Build GRCh38
rs765031703 Chr.6: 8422600-8422606 on Build GRCh38
rs766732814 Chr.6: 8422629-8422629 on Build GRCh38
rs765743302 Chr.6: 8422619-8422619 on Build GRCh38
rs961790317 Chr.6: 8422521-8422521 on Build GRCh38

View Full Product Details