Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 120538792-120539063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNTB1
Entrez Gene ID
6641
Gene Name
syntrophin beta 1
Gene Aliases
59-DAP, A1B, BSYN2, DAPA1B, SNT2, SNT2B1, TIP-43
UniGene
Hs.46701
Gene Chromosome Location
Chr.8: 120535745-120812069 on Build GRCh38
Transcripts
NM_021021.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1187606 c.1555A>G p.I519V Chr.8: 120538939-120538939 on Build GRCh38
4760748 c.1589A>C p.K530T Chr.8: 120538905-120538905 on Build GRCh38
277409 c.1610T>C p.V537A Chr.8: 120538884-120538884 on Build GRCh38
6872567 c.1551G>A p.K517K Chr.8: 120538943-120538943 on Build GRCh38
6472330 c.1533C>T p.D511D Chr.8: 120538961-120538961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779883666 Chr.8: 120538816-120538816 on Build GRCh38
rs751180989 Chr.8: 120538998-120538998 on Build GRCh38
rs200076522 Chr.8: 120538855-120538855 on Build GRCh38
rs760882022 Chr.8: 120538973-120538973 on Build GRCh38
rs777446989 Chr.8: 120538848-120538848 on Build GRCh38
rs765505164 Chr.8: 120538994-120538995 on Build GRCh38
rs551946684 Chr.8: 120539002-120539002 on Build GRCh38
rs760773981 Chr.8: 120538982-120538984 on Build GRCh38
rs761880181 Chr.8: 120538964-120538964 on Build GRCh38
rs377496907 Chr.8: 120538834-120538834 on Build GRCh38
rs375294683 Chr.8: 120539010-120539010 on Build GRCh38
rs888372696 Chr.8: 120539015-120539015 on Build GRCh38
rs767833403 Chr.8: 120538965-120538965 on Build GRCh38
rs766683107 Chr.8: 120538986-120538987 on Build GRCh38
rs141683322 Chr.8: 120538944-120538944 on Build GRCh38
rs779530172 Chr.8: 120538988-120538988 on Build GRCh38
rs879009835 Chr.8: 120538860-120538860 on Build GRCh38
rs1041079731 Chr.8: 120538880-120538880 on Build GRCh38
rs754204085 Chr.8: 120538981-120538981 on Build GRCh38
rs560854527 Chr.8: 120539026-120539026 on Build GRCh38
rs377335826 Chr.8: 120538978-120538978 on Build GRCh38
rs750620687 Chr.8: 120538967-120538967 on Build GRCh38
rs539053079 Chr.8: 120538814-120538814 on Build GRCh38
rs769481277 Chr.8: 120538912-120538912 on Build GRCh38
rs774458076 Chr.8: 120538962-120538962 on Build GRCh38
rs775317055 Chr.8: 120538914-120538914 on Build GRCh38
rs749814893 Chr.8: 120538835-120538835 on Build GRCh38
rs370580771 Chr.8: 120538837-120538837 on Build GRCh38
rs755590242 Chr.8: 120538983-120538983 on Build GRCh38
rs146212838 Chr.8: 120538960-120538960 on Build GRCh38
rs745538942 Chr.8: 120539017-120539017 on Build GRCh38
rs139912456 Chr.8: 120538902-120538902 on Build GRCh38
rs770747477 Chr.8: 120538885-120538885 on Build GRCh38
rs375360963 Chr.8: 120538899-120538899 on Build GRCh38

View Full Product Details