Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88108238-88108463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMYD1
Entrez Gene ID
150572
Gene Name
SET and MYND domain containing 1
Gene Aliases
BOP, KMT3D, ZMYND18, ZMYND22
UniGene
Hs.516176
Gene Chromosome Location
Chr.2: 88067780-88113384 on Build GRCh38
Transcripts
NM_198274.3 XM_005264156.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291384 c.1235G>T p.G412V Chr.2: 88108460-88108460 on Build GRCh38
6484268 c.1189A>T p.M397L Chr.2: 88108414-88108414 on Build GRCh38
5699856 c.1154A>G p.Y385C Chr.2: 88108379-88108379 on Build GRCh38
5426429 c.1215T>C p.H405H Chr.2: 88108440-88108440 on Build GRCh38
3758407 c.1185C>T p.A395A Chr.2: 88108410-88108410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112558914 Chr.2: 88108403-88108403 on Build GRCh38
rs532544199 Chr.2: 88108359-88108359 on Build GRCh38
rs949006276 Chr.2: 88108392-88108392 on Build GRCh38
rs551135161 Chr.2: 88108342-88108342 on Build GRCh38
rs765882434 Chr.2: 88108398-88108398 on Build GRCh38
rs145275251 Chr.2: 88108417-88108417 on Build GRCh38
rs771229558 Chr.2: 88108433-88108433 on Build GRCh38
rs375802266 Chr.2: 88108384-88108384 on Build GRCh38
rs75318533 Chr.2: 88108320-88108320 on Build GRCh38
rs1006194957 Chr.2: 88108281-88108281 on Build GRCh38
rs776538766 Chr.2: 88108434-88108434 on Build GRCh38
rs372431455 Chr.2: 88108368-88108368 on Build GRCh38
rs113530320 Chr.2: 88108410-88108410 on Build GRCh38
rs905198902 Chr.2: 88108263-88108263 on Build GRCh38
rs984659051 Chr.2: 88108336-88108336 on Build GRCh38
rs1028339604 Chr.2: 88108319-88108319 on Build GRCh38
rs753069207 Chr.2: 88108399-88108399 on Build GRCh38
rs1008227652 Chr.2: 88108400-88108400 on Build GRCh38
rs145939300 Chr.2: 88108405-88108405 on Build GRCh38
rs763779676 Chr.2: 88108404-88108404 on Build GRCh38
rs759933899 Chr.2: 88108383-88108383 on Build GRCh38
rs747181609 Chr.2: 88108431-88108431 on Build GRCh38
rs146456865 Chr.2: 88108418-88108418 on Build GRCh38
rs200376153 Chr.2: 88108441-88108441 on Build GRCh38
rs772246016 Chr.2: 88108329-88108329 on Build GRCh38
rs776826023 Chr.2: 88108377-88108377 on Build GRCh38
rs139345589 Chr.2: 88108411-88108411 on Build GRCh38
rs753426741 Chr.2: 88108342-88108343 on Build GRCh38
rs867439931 Chr.2: 88108409-88108409 on Build GRCh38
rs763342067 Chr.2: 88108444-88108444 on Build GRCh38
rs370398156 Chr.2: 88108375-88108375 on Build GRCh38
rs148646168 Chr.2: 88108352-88108352 on Build GRCh38
rs2970918 Chr.2: 88108440-88108440 on Build GRCh38
rs780000342 Chr.2: 88108413-88108413 on Build GRCh38
rs777909972 Chr.2: 88108420-88108420 on Build GRCh38

View Full Product Details