Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155047626-155047885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCST1
Entrez Gene ID
149095
Gene Name
DC-STAMP domain containing 1
Gene Aliases
-
UniGene
Hs.567717
Gene Chromosome Location
Chr.1: 155033806-155050930 on Build GRCh38
Transcripts
NM_001143687.2 NM_152494.3 NR_040772.1 NR_040773.1

Gene Symbol
LOC100505666
Entrez Gene ID
100505666
Gene Name
uncharacterized LOC100505666
Gene Aliases
-
UniGene
Hs.657533
Gene Chromosome Location
Chr.1: 155045191-155063991 on Build GRCh38
Transcripts
NM_001143687.2 NM_152494.3 NR_040772.1 NR_040773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4676595 c.1671G>A p.P557P Chr.1: 155047845-155047845 on Build GRCh38
4022926 c.1635C>T p.G545G Chr.1: 155047809-155047809 on Build GRCh38
1743152 c.1704G>C p.Q568H Chr.1: 155047878-155047878 on Build GRCh38
4676596 c.1671G>A p.P557P Chr.1: 155047845-155047845 on Build GRCh38
4676593 c.1667T>C p.V556A Chr.1: 155047841-155047841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745861868 Chr.1: 155047838-155047838 on Build GRCh38
rs766931391 Chr.1: 155047759-155047759 on Build GRCh38
rs777671882 Chr.1: 155047802-155047802 on Build GRCh38
rs559328902 Chr.1: 155047662-155047662 on Build GRCh38
rs890993729 Chr.1: 155047778-155047778 on Build GRCh38
rs899609235 Chr.1: 155047694-155047694 on Build GRCh38
rs372164098 Chr.1: 155047736-155047736 on Build GRCh38
rs542258929 Chr.1: 155047646-155047646 on Build GRCh38
rs199907649 Chr.1: 155047787-155047787 on Build GRCh38
rs773516092 Chr.1: 155047849-155047849 on Build GRCh38
rs766728250 Chr.1: 155047858-155047858 on Build GRCh38
rs761988652 Chr.1: 155047845-155047845 on Build GRCh38
rs377078307 Chr.1: 155047744-155047744 on Build GRCh38
rs997055020 Chr.1: 155047722-155047722 on Build GRCh38
rs1003392192 Chr.1: 155047857-155047857 on Build GRCh38
rs375311605 Chr.1: 155047786-155047786 on Build GRCh38
rs144291978 Chr.1: 155047847-155047847 on Build GRCh38
rs544553710 Chr.1: 155047806-155047806 on Build GRCh38
rs768771276 Chr.1: 155047843-155047843 on Build GRCh38
rs776738245 Chr.1: 155047844-155047844 on Build GRCh38
rs770719792 Chr.1: 155047821-155047821 on Build GRCh38
rs563114037 Chr.1: 155047856-155047856 on Build GRCh38
rs755873246 Chr.1: 155047794-155047794 on Build GRCh38
rs763389865 Chr.1: 155047851-155047851 on Build GRCh38
rs891835230 Chr.1: 155047701-155047701 on Build GRCh38
rs980895168 Chr.1: 155047763-155047763 on Build GRCh38
rs764371110 Chr.1: 155047793-155047794 on Build GRCh38
rs528457705 Chr.1: 155047808-155047808 on Build GRCh38
rs749118784 Chr.1: 155047804-155047804 on Build GRCh38
rs747782403 Chr.1: 155047791-155047791 on Build GRCh38
rs774354116 Chr.1: 155047826-155047826 on Build GRCh38
rs753940821 Chr.1: 155047847-155047849 on Build GRCh38
rs1054456492 Chr.1: 155047652-155047652 on Build GRCh38

View Full Product Details