Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 67007996-67008239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KBTBD8
Entrez Gene ID
84541
Gene Name
kelch repeat and BTB domain containing 8
Gene Aliases
TA-KRP, TAKRP
UniGene
Hs.116665
Gene Chromosome Location
Chr.3: 66998303-67011210 on Build GRCh38
Transcripts
NM_032505.2 XM_005264771.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848085 c.1454G>T p.R485L Chr.3: 67008111-67008111 on Build GRCh38
377927 c.1473T>C p.C491C Chr.3: 67008130-67008130 on Build GRCh38
3597186 c.1481C>T p.S494F Chr.3: 67008138-67008138 on Build GRCh38
75068 c.1513C>G p.Q505E Chr.3: 67008170-67008170 on Build GRCh38
194567 c.1567G>A p.V523I Chr.3: 67008224-67008224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755394459 Chr.3: 67008191-67008191 on Build GRCh38
rs142381130 Chr.3: 67008169-67008169 on Build GRCh38
rs534443174 Chr.3: 67008218-67008218 on Build GRCh38
rs530554608 Chr.3: 67008195-67008195 on Build GRCh38
rs946756795 Chr.3: 67008164-67008164 on Build GRCh38
rs748888735 Chr.3: 67008189-67008189 on Build GRCh38
rs565525319 Chr.3: 67008100-67008100 on Build GRCh38
rs145930523 Chr.3: 67008064-67008064 on Build GRCh38
rs763644100 Chr.3: 67008124-67008124 on Build GRCh38
rs377495210 Chr.3: 67008065-67008065 on Build GRCh38
rs779539393 Chr.3: 67008038-67008038 on Build GRCh38
rs779778192 Chr.3: 67008178-67008178 on Build GRCh38
rs749700317 Chr.3: 67008048-67008048 on Build GRCh38
rs757253028 Chr.3: 67008157-67008157 on Build GRCh38
rs143301489 Chr.3: 67008107-67008107 on Build GRCh38
rs753010384 Chr.3: 67008025-67008026 on Build GRCh38
rs763821679 Chr.3: 67008126-67008126 on Build GRCh38
rs779484880 Chr.3: 67008194-67008194 on Build GRCh38
rs749989300 Chr.3: 67008160-67008160 on Build GRCh38
rs766667457 Chr.3: 67008110-67008110 on Build GRCh38
rs202223775 Chr.3: 67008111-67008111 on Build GRCh38
rs912816486 Chr.3: 67008033-67008033 on Build GRCh38
rs367559555 Chr.3: 67008158-67008158 on Build GRCh38
rs773403096 Chr.3: 67008086-67008086 on Build GRCh38
rs951097655 Chr.3: 67008061-67008061 on Build GRCh38
rs551365299 Chr.3: 67008057-67008057 on Build GRCh38
rs201286846 Chr.3: 67008152-67008152 on Build GRCh38
rs775008928 Chr.3: 67008062-67008062 on Build GRCh38
rs944350795 Chr.3: 67008063-67008063 on Build GRCh38

View Full Product Details