Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65664351-65664582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4A
Entrez Gene ID
10260
Gene Name
DENN domain containing 4A
Gene Aliases
IRLB, MYCPBP
UniGene
Hs.654567
Gene Chromosome Location
Chr.15: 65659123-65792293 on Build GRCh38
Transcripts
NM_001144823.2 NM_001320835.1 NM_005848.4 XM_005254120.4 XM_005254121.3 XM_005254122.4 XM_006720371.3 XM_011521157.2 XM_017021862.1 XM_017021863.1 XM_017021865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6406798 c.5383A>T p.R1795* Chr.15: 65664567-65664567 on Build GRCh38
964107 c.5404G>T p.E1802* Chr.15: 65664381-65664381 on Build GRCh38
1588043 c.5533G>T p.E1845* Chr.15: 65664381-65664381 on Build GRCh38
4056336 c.5520-2A>G p.? Chr.15: 65664396-65664396 on Build GRCh38
4056337 c.5391-2A>G p.? Chr.15: 65664396-65664396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953466057 Chr.15: 65664478-65664478 on Build GRCh38
rs984855035 Chr.15: 65664492-65664492 on Build GRCh38
rs774347121 Chr.15: 65664549-65664549 on Build GRCh38
rs770072745 Chr.15: 65664535-65664535 on Build GRCh38
rs752545903 Chr.15: 65664413-65664413 on Build GRCh38
rs773579883 Chr.15: 65664537-65664537 on Build GRCh38
rs771159415 Chr.15: 65664548-65664550 on Build GRCh38
rs115822209 Chr.15: 65664514-65664514 on Build GRCh38
rs759435120 Chr.15: 65664393-65664393 on Build GRCh38
rs201513707 Chr.15: 65664540-65664540 on Build GRCh38
rs749548954 Chr.15: 65664534-65664535 on Build GRCh38
rs940714248 Chr.15: 65664461-65664461 on Build GRCh38
rs374936047 Chr.15: 65664532-65664532 on Build GRCh38
rs1029432963 Chr.15: 65664398-65664398 on Build GRCh38
rs777800018 Chr.15: 65664403-65664404 on Build GRCh38
rs777408080 Chr.15: 65664444-65664444 on Build GRCh38
rs769092724 Chr.15: 65664529-65664529 on Build GRCh38
rs369246257 Chr.15: 65664541-65664541 on Build GRCh38
rs755760712 Chr.15: 65664441-65664441 on Build GRCh38
rs368434376 Chr.15: 65664523-65664523 on Build GRCh38
rs563444672 Chr.15: 65664405-65664405 on Build GRCh38
rs997514130 Chr.15: 65664379-65664379 on Build GRCh38
rs780384012 Chr.15: 65664520-65664520 on Build GRCh38
rs372297633 Chr.15: 65664410-65664410 on Build GRCh38
rs753289592 Chr.15: 65664512-65664512 on Build GRCh38

View Full Product Details