Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38403137-38403398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98C
Entrez Gene ID
147965
Gene Name
family with sequence similarity 98 member C
Gene Aliases
-
UniGene
Hs.355162
Gene Chromosome Location
Chr.19: 38403124-38409088 on Build GRCh38
Transcripts
NM_174905.3 XM_017026354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
418289 c.15G>A p.K5K Chr.19: 38403168-38403168 on Build GRCh38
4508872 c.78C>A p.V26V Chr.19: 38403350-38403350 on Build GRCh38
3766332 c.63G>C p.L21L Chr.19: 38403216-38403216 on Build GRCh38
4639040 c.24G>A p.A8A Chr.19: 38403177-38403177 on Build GRCh38
6028197 c.30delG p.A12fs*56 Chr.19: 38403183-38403183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751386424 Chr.19: 38403176-38403176 on Build GRCh38
rs771487369 Chr.19: 38403198-38403198 on Build GRCh38
rs763730833 Chr.19: 38403170-38403170 on Build GRCh38
rs576614116 Chr.19: 38403357-38403357 on Build GRCh38
rs3745957 Chr.19: 38403216-38403216 on Build GRCh38
rs3745959 Chr.19: 38403295-38403295 on Build GRCh38
rs868311826 Chr.19: 38403336-38403336 on Build GRCh38
rs867495583 Chr.19: 38403247-38403247 on Build GRCh38
rs767257437 Chr.19: 38403349-38403349 on Build GRCh38
rs939913770 Chr.19: 38403199-38403199 on Build GRCh38
rs772992244 Chr.19: 38403350-38403350 on Build GRCh38
rs867040571 Chr.19: 38403318-38403318 on Build GRCh38
rs761567539 Chr.19: 38403239-38403239 on Build GRCh38
rs71355001 Chr.19: 38403160-38403160 on Build GRCh38
rs778336998 Chr.19: 38403276-38403276 on Build GRCh38
rs888851848 Chr.19: 38403290-38403290 on Build GRCh38
rs761759274 Chr.19: 38403344-38403344 on Build GRCh38
rs777941272 Chr.19: 38403195-38403195 on Build GRCh38
rs776960731 Chr.19: 38403200-38403200 on Build GRCh38
rs375145477 Chr.19: 38403221-38403221 on Build GRCh38
rs750119042 Chr.19: 38403186-38403186 on Build GRCh38
rs757105888 Chr.19: 38403177-38403177 on Build GRCh38
rs937865896 Chr.19: 38403222-38403222 on Build GRCh38
rs746020377 Chr.19: 38403304-38403304 on Build GRCh38
rs760259240 Chr.19: 38403376-38403376 on Build GRCh38
rs774325753 Chr.19: 38403331-38403331 on Build GRCh38
rs753401549 Chr.19: 38403262-38403262 on Build GRCh38
rs766943368 Chr.19: 38403349-38403349 on Build GRCh38
rs761036793 Chr.19: 38403266-38403267 on Build GRCh38
rs571712703 Chr.19: 38403257-38403257 on Build GRCh38
rs750026459 Chr.19: 38403256-38403256 on Build GRCh38
rs779559971 Chr.19: 38403189-38403189 on Build GRCh38
rs767345479 Chr.19: 38403249-38403249 on Build GRCh38
rs1027753656 Chr.19: 38403175-38403175 on Build GRCh38
rs754503966 Chr.19: 38403269-38403269 on Build GRCh38
rs201469183 Chr.19: 38403192-38403192 on Build GRCh38
rs867365579 Chr.19: 38403342-38403342 on Build GRCh38
rs757648183 Chr.19: 38403285-38403285 on Build GRCh38
rs946481219 Chr.19: 38403294-38403294 on Build GRCh38
rs767623313 Chr.19: 38403233-38403241 on Build GRCh38
rs752530256 Chr.19: 38403162-38403162 on Build GRCh38
rs1055253388 Chr.19: 38403261-38403261 on Build GRCh38
rs537029653 Chr.19: 38403267-38403267 on Build GRCh38
rs780820374 Chr.19: 38403178-38403178 on Build GRCh38
rs3745958 Chr.19: 38403284-38403284 on Build GRCh38
rs751283524 Chr.19: 38403351-38403351 on Build GRCh38
rs542648656 Chr.19: 38403308-38403308 on Build GRCh38
rs191201342 Chr.19: 38403306-38403306 on Build GRCh38
rs993377375 Chr.19: 38403313-38403313 on Build GRCh38
rs3745960 Chr.19: 38403310-38403310 on Build GRCh38
rs762980062 Chr.19: 38403166-38403166 on Build GRCh38
rs759861575 Chr.19: 38403214-38403214 on Build GRCh38
rs764083918 Chr.19: 38403231-38403231 on Build GRCh38
rs751521464 Chr.19: 38403234-38403234 on Build GRCh38
rs747385715 Chr.19: 38403197-38403197 on Build GRCh38

View Full Product Details