Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 12901218-12901448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQSEC1
Entrez Gene ID
9922
Gene Name
IQ motif and Sec7 domain 1
Gene Aliases
ARF-GEP100, ARFGEP100, BRAG2, GEP100
UniGene
Hs.475506
Gene Chromosome Location
Chr.3: 12897043-13282998 on Build GRCh38
Transcripts
NM_001134382.2 NM_014869.6 XM_011534304.2 XM_011534305.2 XM_011534306.2 XM_011534307.2 XM_011534308.2 XM_011534310.2 XM_011534311.2 XM_011534312.2 XM_011534313.2 XM_011534314.2 XM_011534315.2 XM_017007581.1 XM_017007582.1

SNP Detail

SNP ID Chromosome Location
rs758422581 Chr.3: 12901375-12901375 on Build GRCh38
rs752070965 Chr.3: 12901379-12901379 on Build GRCh38
rs763819248 Chr.3: 12901255-12901255 on Build GRCh38
rs755482053 Chr.3: 12901390-12901390 on Build GRCh38
rs747490315 Chr.3: 12901268-12901268 on Build GRCh38
rs375583511 Chr.3: 12901250-12901250 on Build GRCh38
rs760462253 Chr.3: 12901252-12901252 on Build GRCh38
rs750655631 Chr.3: 12901369-12901369 on Build GRCh38
rs545063086 Chr.3: 12901408-12901408 on Build GRCh38
rs781447591 Chr.3: 12901274-12901274 on Build GRCh38
rs771724385 Chr.3: 12901300-12901300 on Build GRCh38
rs137962390 Chr.3: 12901259-12901261 on Build GRCh38
rs759052806 Chr.3: 12901296-12901296 on Build GRCh38
rs542687791 Chr.3: 12901334-12901334 on Build GRCh38
rs866930156 Chr.3: 12901377-12901377 on Build GRCh38
rs913276765 Chr.3: 12901317-12901317 on Build GRCh38
rs565809901 Chr.3: 12901256-12901256 on Build GRCh38
rs202035528 Chr.3: 12901378-12901378 on Build GRCh38
rs200289066 Chr.3: 12901407-12901407 on Build GRCh38
rs914723275 Chr.3: 12901359-12901359 on Build GRCh38
rs1016201679 Chr.3: 12901258-12901258 on Build GRCh38
rs565142672 Chr.3: 12901409-12901409 on Build GRCh38
rs397988742 Chr.3: 12901352-12901352 on Build GRCh38
rs866274367 Chr.3: 12901333-12901333 on Build GRCh38
rs557020896 Chr.3: 12901275-12901275 on Build GRCh38
rs780006602 Chr.3: 12901349-12901349 on Build GRCh38
rs573868764 Chr.3: 12901289-12901289 on Build GRCh38
rs768743841 Chr.3: 12901353-12901354 on Build GRCh38
rs552924984 Chr.3: 12901360-12901360 on Build GRCh38
rs778950868 Chr.3: 12901260-12901260 on Build GRCh38
rs866819128 Chr.3: 12901349-12901349 on Build GRCh38
rs866702107 Chr.3: 12901341-12901341 on Build GRCh38
rs760563114 Chr.3: 12901343-12901343 on Build GRCh38
rs932144204 Chr.3: 12901420-12901420 on Build GRCh38
rs537355221 Chr.3: 12901269-12901269 on Build GRCh38
rs1002911492 Chr.3: 12901400-12901400 on Build GRCh38
rs534679416 Chr.3: 12901259-12901259 on Build GRCh38
rs753626236 Chr.3: 12901346-12901346 on Build GRCh38
rs571426372 Chr.3: 12901262-12901262 on Build GRCh38
rs902887326 Chr.3: 12901391-12901391 on Build GRCh38
rs763918726 Chr.3: 12901345-12901345 on Build GRCh38
rs1013983154 Chr.3: 12901253-12901253 on Build GRCh38
rs951459571 Chr.3: 12901265-12901265 on Build GRCh38
rs770769839 Chr.3: 12901285-12901285 on Build GRCh38
rs185165059 Chr.3: 12901261-12901261 on Build GRCh38

View Full Product Details