Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70400016-70400265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNMB4
Entrez Gene ID
27345
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 4
Gene Aliases
-
UniGene
Hs.525529
Gene Chromosome Location
Chr.12: 70366220-70434292 on Build GRCh38
Transcripts
NM_014505.5 XM_011538188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4805988 c.392T>C p.M131T Chr.12: 70400264-70400264 on Build GRCh38
6073721 c.391A>G p.M131V Chr.12: 70400263-70400263 on Build GRCh38
3664874 c.347T>C p.I116T Chr.12: 70400219-70400219 on Build GRCh38
4044588 c.385A>G p.S129G Chr.12: 70400257-70400257 on Build GRCh38
6481528 c.393G>C p.M131I Chr.12: 70400265-70400265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756214838 Chr.12: 70400110-70400110 on Build GRCh38
rs943887911 Chr.12: 70400146-70400146 on Build GRCh38
rs867009582 Chr.12: 70400101-70400101 on Build GRCh38
rs935258947 Chr.12: 70400141-70400141 on Build GRCh38
rs977073435 Chr.12: 70400066-70400066 on Build GRCh38
rs149071110 Chr.12: 70400042-70400042 on Build GRCh38
rs912358911 Chr.12: 70400112-70400112 on Build GRCh38
rs112314761 Chr.12: 70400206-70400206 on Build GRCh38
rs1042202943 Chr.12: 70400096-70400096 on Build GRCh38
rs752810846 Chr.12: 70400074-70400074 on Build GRCh38
rs781226845 Chr.12: 70400180-70400180 on Build GRCh38
rs748009617 Chr.12: 70400225-70400225 on Build GRCh38
rs1052191996 Chr.12: 70400148-70400148 on Build GRCh38
rs571386810 Chr.12: 70400167-70400167 on Build GRCh38
rs947440796 Chr.12: 70400194-70400194 on Build GRCh38
rs1039569419 Chr.12: 70400162-70400162 on Build GRCh38
rs201392498 Chr.12: 70400164-70400164 on Build GRCh38

View Full Product Details