Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149715949-149716194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEA11
Entrez Gene ID
4110
Gene Name
MAGE family member A11
Gene Aliases
CT1.11, MAGE-11, MAGE11, MAGEA-11
UniGene
Hs.670252
Gene Chromosome Location
Chr.X: 149688202-149717268 on Build GRCh38
Transcripts
NM_001011544.1 NM_005366.4 XM_011531164.2 XM_017029521.1 XM_017029522.1

SNP Detail

SNP ID Chromosome Location
rs902243994 Chr.X: 149716133-149716133 on Build GRCh38
rs373850281 Chr.X: 149716045-149716045 on Build GRCh38
rs782664058 Chr.X: 149715973-149715973 on Build GRCh38
rs782156874 Chr.X: 149716109-149716109 on Build GRCh38
rs782315137 Chr.X: 149716037-149716037 on Build GRCh38
rs782535658 Chr.X: 149716132-149716132 on Build GRCh38
rs781826740 Chr.X: 149716101-149716101 on Build GRCh38
rs782234120 Chr.X: 149716162-149716162 on Build GRCh38
rs781855717 Chr.X: 149715982-149715982 on Build GRCh38
rs782580746 Chr.X: 149716039-149716039 on Build GRCh38
rs207479030 Chr.X: 149715986-149715986 on Build GRCh38
rs782778956 Chr.X: 149716096-149716096 on Build GRCh38
rs781813293 Chr.X: 149716048-149716048 on Build GRCh38
rs374038807 Chr.X: 149716128-149716128 on Build GRCh38
rs782521042 Chr.X: 149715991-149715991 on Build GRCh38
rs781883773 Chr.X: 149716071-149716071 on Build GRCh38
rs782226278 Chr.X: 149716030-149716030 on Build GRCh38
rs370004708 Chr.X: 149715994-149715994 on Build GRCh38
rs370362756 Chr.X: 149716067-149716067 on Build GRCh38
rs782565753 Chr.X: 149716110-149716110 on Build GRCh38
rs200719649 Chr.X: 149716053-149716053 on Build GRCh38
rs782337606 Chr.X: 149716058-149716058 on Build GRCh38
rs782472807 Chr.X: 149716051-149716051 on Build GRCh38
rs782126718 Chr.X: 149716055-149716055 on Build GRCh38

View Full Product Details