Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101716639-101716904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPT1
Entrez Gene ID
56994
Gene Name
choline phosphotransferase 1
Gene Aliases
CPT, CPT1
UniGene
Hs.293077
Gene Chromosome Location
Chr.12: 101697639-101752038 on Build GRCh38
Transcripts
NM_020244.2 XM_011538574.1 XM_011538575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6256070 c.567G>T p.V189V Chr.12: 101716731-101716731 on Build GRCh38
1706129 c.607G>C p.V203L Chr.12: 101716771-101716771 on Build GRCh38
3703809 c.647C>T p.T216M Chr.12: 101716811-101716811 on Build GRCh38
1677287 c.583C>T p.Q195* Chr.12: 101716747-101716747 on Build GRCh38
5628018 c.640G>A p.D214N Chr.12: 101716804-101716804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774438855 Chr.12: 101716754-101716754 on Build GRCh38
rs746839908 Chr.12: 101716833-101716833 on Build GRCh38
rs747000998 Chr.12: 101716706-101716706 on Build GRCh38
rs773239386 Chr.12: 101716731-101716731 on Build GRCh38
rs369207872 Chr.12: 101716702-101716702 on Build GRCh38
rs756891939 Chr.12: 101716677-101716677 on Build GRCh38
rs1037375264 Chr.12: 101716732-101716732 on Build GRCh38
rs369936908 Chr.12: 101716793-101716793 on Build GRCh38
rs764728188 Chr.12: 101716795-101716795 on Build GRCh38
rs771921270 Chr.12: 101716699-101716699 on Build GRCh38
rs2303627 Chr.12: 101716835-101716835 on Build GRCh38
rs199582822 Chr.12: 101716812-101716812 on Build GRCh38
rs758168629 Chr.12: 101716813-101716813 on Build GRCh38
rs760928430 Chr.12: 101716752-101716752 on Build GRCh38
rs373986667 Chr.12: 101716697-101716697 on Build GRCh38
rs749319220 Chr.12: 101716853-101716853 on Build GRCh38
rs139763457 Chr.12: 101716776-101716776 on Build GRCh38
rs761305017 Chr.12: 101716794-101716794 on Build GRCh38
rs866358127 Chr.12: 101716688-101716688 on Build GRCh38
rs779867509 Chr.12: 101716825-101716825 on Build GRCh38
rs146419734 Chr.12: 101716698-101716698 on Build GRCh38
rs764940198 Chr.12: 101716811-101716811 on Build GRCh38
rs368509107 Chr.12: 101716847-101716847 on Build GRCh38
rs771123242 Chr.12: 101716745-101716745 on Build GRCh38
rs959247964 Chr.12: 101716766-101716766 on Build GRCh38
rs753338365 Chr.12: 101716798-101716798 on Build GRCh38
rs149385670 Chr.12: 101716792-101716792 on Build GRCh38
rs778346709 Chr.12: 101716795-101716809 on Build GRCh38
rs779954959 Chr.12: 101716704-101716704 on Build GRCh38
rs1034918202 Chr.12: 101716727-101716727 on Build GRCh38
rs377435526 Chr.12: 101716723-101716723 on Build GRCh38
rs759936738 Chr.12: 101716768-101716768 on Build GRCh38
rs372662246 Chr.12: 101716722-101716722 on Build GRCh38
rs749353294 Chr.12: 101716739-101716739 on Build GRCh38
rs756930625 Chr.12: 101716772-101716772 on Build GRCh38
rs1046639237 Chr.12: 101716819-101716819 on Build GRCh38
rs756725928 Chr.12: 101716799-101716799 on Build GRCh38

View Full Product Details