Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27964811-27965070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDX2
Entrez Gene ID
1045
Gene Name
caudal type homeobox 2
Gene Aliases
CDX-3, CDX2/AS, CDX3
UniGene
Hs.127383
Gene Chromosome Location
Chr.13: 27960918-27969368 on Build GRCh38
Transcripts
NM_001265.4 XM_011534875.2 XM_011534876.2 XM_011534877.2 XM_011534878.2 XM_011534879.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5036696 c.604G>C p.E202Q Chr.13: 27964953-27964953 on Build GRCh38
5897054 c.583G>A p.D195N Chr.13: 27964974-27964974 on Build GRCh38
381912 c.597_598GG>TT p.L199>? Chr.13: 27964959-27964960 on Build GRCh38
946459 c.661G>A p.A221T Chr.13: 27964896-27964896 on Build GRCh38
3468046 c.600G>A p.E200E Chr.13: 27964957-27964957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747360741 Chr.13: 27964897-27964897 on Build GRCh38
rs564888160 Chr.13: 27964892-27964892 on Build GRCh38
rs775395145 Chr.13: 27964921-27964921 on Build GRCh38
rs190770109 Chr.13: 27964842-27964842 on Build GRCh38
rs867906635 Chr.13: 27965021-27965021 on Build GRCh38
rs756121674 Chr.13: 27964933-27964933 on Build GRCh38
rs762169087 Chr.13: 27964832-27964832 on Build GRCh38
rs149977259 Chr.13: 27964932-27964932 on Build GRCh38
rs747590531 Chr.13: 27965002-27965002 on Build GRCh38
rs781315595 Chr.13: 27964862-27964862 on Build GRCh38
rs757964058 Chr.13: 27965017-27965017 on Build GRCh38
rs769882734 Chr.13: 27965040-27965040 on Build GRCh38
rs771445196 Chr.13: 27965034-27965034 on Build GRCh38
rs913721335 Chr.13: 27964913-27964913 on Build GRCh38
rs386769409 Chr.13: 27965048-27965049 on Build GRCh38
rs765166922 Chr.13: 27964858-27964858 on Build GRCh38
rs771353023 Chr.13: 27964900-27964900 on Build GRCh38
rs886131493 Chr.13: 27964975-27964975 on Build GRCh38
rs746279056 Chr.13: 27965038-27965038 on Build GRCh38
rs773916303 Chr.13: 27964831-27964831 on Build GRCh38
rs765521027 Chr.13: 27964917-27964917 on Build GRCh38
rs754544806 Chr.13: 27964954-27964954 on Build GRCh38
rs757097084 Chr.13: 27964861-27964861 on Build GRCh38
rs754672457 Chr.13: 27964886-27964886 on Build GRCh38
rs763139212 Chr.13: 27964931-27964931 on Build GRCh38
rs201884582 Chr.13: 27964843-27964843 on Build GRCh38
rs376260625 Chr.13: 27964945-27964945 on Build GRCh38
rs2481953 Chr.13: 27965048-27965048 on Build GRCh38
rs901765967 Chr.13: 27965039-27965039 on Build GRCh38
rs111883191 Chr.13: 27964847-27964847 on Build GRCh38
rs377404719 Chr.13: 27964854-27964854 on Build GRCh38
rs562390909 Chr.13: 27964906-27964906 on Build GRCh38
rs371213778 Chr.13: 27964860-27964860 on Build GRCh38
rs879439886 Chr.13: 27965048-27965049 on Build GRCh38
rs778236024 Chr.13: 27964891-27964891 on Build GRCh38
rs1029665012 Chr.13: 27964850-27964850 on Build GRCh38
rs764199114 Chr.13: 27965051-27965051 on Build GRCh38
rs759623152 Chr.13: 27964916-27964916 on Build GRCh38
rs773759361 Chr.13: 27964896-27964896 on Build GRCh38
rs930415507 Chr.13: 27964907-27964907 on Build GRCh38
rs368588724 Chr.13: 27965028-27965028 on Build GRCh38
rs1004975547 Chr.13: 27965001-27965001 on Build GRCh38
rs370757560 Chr.13: 27964846-27964846 on Build GRCh38
rs756026762 Chr.13: 27964863-27964863 on Build GRCh38
rs201820089 Chr.13: 27964868-27964868 on Build GRCh38
rs887290682 Chr.13: 27964839-27964839 on Build GRCh38
rs761070555 Chr.13: 27964844-27964844 on Build GRCh38
rs3742110 Chr.13: 27965049-27965049 on Build GRCh38
rs778666045 Chr.13: 27964964-27964964 on Build GRCh38
rs868166000 Chr.13: 27965022-27965022 on Build GRCh38
rs779845244 Chr.13: 27964864-27964864 on Build GRCh38
rs778248122 Chr.13: 27965024-27965024 on Build GRCh38
rs753689670 Chr.13: 27964948-27964948 on Build GRCh38
rs1028817287 Chr.13: 27965041-27965041 on Build GRCh38
rs201267204 Chr.13: 27965035-27965035 on Build GRCh38

View Full Product Details