Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48115810-48116053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIG1
Entrez Gene ID
3978
Gene Name
DNA ligase 1
Gene Aliases
-
UniGene
Hs.1770
Gene Chromosome Location
Chr.19: 48115445-48170603 on Build GRCh38
Transcripts
NM_000234.2 NM_001289063.1 NM_001289064.1 NM_001320970.1 NM_001320971.1 NR_110296.1 NR_135497.1 NR_135498.1 NR_135499.1 NR_135500.1 NR_135501.1 XM_017026809.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751024 c.2600G>A p.G867D Chr.19: 48115949-48115949 on Build GRCh38
998857 c.2651C>T p.P884L Chr.19: 48115898-48115898 on Build GRCh38
5942832 c.2611C>T p.R871C Chr.19: 48115938-48115938 on Build GRCh38
7332216 c.2604C>T p.I868I Chr.19: 48115945-48115945 on Build GRCh38
275855 c.2629C>T p.R877* Chr.19: 48115920-48115920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751739483 Chr.19: 48115846-48115846 on Build GRCh38
rs776608940 Chr.19: 48115928-48115928 on Build GRCh38
rs142452745 Chr.19: 48115992-48115992 on Build GRCh38
rs748252507 Chr.19: 48115884-48115884 on Build GRCh38
rs11666150 Chr.19: 48115873-48115873 on Build GRCh38
rs1048821455 Chr.19: 48115847-48115847 on Build GRCh38
rs765288678 Chr.19: 48115937-48115937 on Build GRCh38
rs977323232 Chr.19: 48116029-48116029 on Build GRCh38
rs778962963 Chr.19: 48115845-48115845 on Build GRCh38
rs746305040 Chr.19: 48116006-48116006 on Build GRCh38
rs778165966 Chr.19: 48115896-48115896 on Build GRCh38
rs757330874 Chr.19: 48115844-48115844 on Build GRCh38
rs753676820 Chr.19: 48115842-48115842 on Build GRCh38
rs35100567 Chr.19: 48115897-48115897 on Build GRCh38
rs745577016 Chr.19: 48116009-48116009 on Build GRCh38
rs763835099 Chr.19: 48115923-48115923 on Build GRCh38
rs182766545 Chr.19: 48115975-48115975 on Build GRCh38
rs749699872 Chr.19: 48115993-48115993 on Build GRCh38
rs760305891 Chr.19: 48115920-48115920 on Build GRCh38
rs1051793362 Chr.19: 48115914-48115914 on Build GRCh38
rs756514492 Chr.19: 48115887-48115887 on Build GRCh38
rs774814943 Chr.19: 48115900-48115900 on Build GRCh38
rs898652788 Chr.19: 48115964-48115964 on Build GRCh38
rs146892854 Chr.19: 48116007-48116007 on Build GRCh38
rs201176712 Chr.19: 48115898-48115898 on Build GRCh38
rs767523594 Chr.19: 48116008-48116009 on Build GRCh38
rs928616253 Chr.19: 48116018-48116018 on Build GRCh38
rs761679526 Chr.19: 48115932-48115932 on Build GRCh38
rs761865488 Chr.19: 48115961-48115961 on Build GRCh38
rs745417422 Chr.19: 48115904-48115904 on Build GRCh38
rs781506154 Chr.19: 48115883-48115883 on Build GRCh38
rs376541305 Chr.19: 48115984-48115984 on Build GRCh38
rs973375101 Chr.19: 48116028-48116028 on Build GRCh38
rs200192992 Chr.19: 48115840-48115840 on Build GRCh38
rs775226298 Chr.19: 48115919-48115919 on Build GRCh38
rs1037193058 Chr.19: 48115961-48115961 on Build GRCh38
rs971374104 Chr.19: 48116019-48116019 on Build GRCh38
rs750286209 Chr.19: 48115960-48115960 on Build GRCh38
rs756391107 Chr.19: 48115976-48115976 on Build GRCh38
rs771419079 Chr.19: 48115913-48115913 on Build GRCh38
rs199958887 Chr.19: 48115969-48115969 on Build GRCh38
rs757565812 Chr.19: 48116000-48116000 on Build GRCh38
rs534721657 Chr.19: 48115970-48115970 on Build GRCh38

View Full Product Details