Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 215586522-215586777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD3
Entrez Gene ID
51133
Gene Name
potassium channel tetramerization domain containing 3
Gene Aliases
NY-REN-45
UniGene
Hs.335139
Gene Chromosome Location
Chr.1: 215567185-215621821 on Build GRCh38
Transcripts
NM_001319294.1 NM_001319295.1 NM_016121.4 XM_005273158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6061308 c.663G>C p.T221T Chr.1: 215586531-215586531 on Build GRCh38
678648 c.739G>A p.D247N Chr.1: 215586607-215586607 on Build GRCh38
425287 c.725C>T p.P242L Chr.1: 215586593-215586593 on Build GRCh38
5448411 c.696A>G p.V232V Chr.1: 215586564-215586564 on Build GRCh38
7201589 c.688G>C p.E230Q Chr.1: 215586556-215586556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772880495 Chr.1: 215586624-215586624 on Build GRCh38
rs769618795 Chr.1: 215586602-215586602 on Build GRCh38
rs778360428 Chr.1: 215586596-215586596 on Build GRCh38
rs774553049 Chr.1: 215586664-215586664 on Build GRCh38
rs202078864 Chr.1: 215586715-215586715 on Build GRCh38
rs952570240 Chr.1: 215586700-215586700 on Build GRCh38
rs747704075 Chr.1: 215586597-215586597 on Build GRCh38
rs756345486 Chr.1: 215586705-215586705 on Build GRCh38
rs367892719 Chr.1: 215586689-215586689 on Build GRCh38
rs754972599 Chr.1: 215586588-215586588 on Build GRCh38
rs368987741 Chr.1: 215586723-215586723 on Build GRCh38
rs767528806 Chr.1: 215586667-215586667 on Build GRCh38
rs1002990070 Chr.1: 215586555-215586555 on Build GRCh38
rs771502821 Chr.1: 215586622-215586622 on Build GRCh38
rs768619362 Chr.1: 215586746-215586746 on Build GRCh38
rs985298953 Chr.1: 215586718-215586718 on Build GRCh38
rs753671676 Chr.1: 215586710-215586710 on Build GRCh38
rs376812729 Chr.1: 215586722-215586722 on Build GRCh38
rs764738821 Chr.1: 215586663-215586663 on Build GRCh38
rs779723592 Chr.1: 215586550-215586550 on Build GRCh38
rs770139728 Chr.1: 215586631-215586631 on Build GRCh38
rs149751728 Chr.1: 215586627-215586627 on Build GRCh38
rs969826329 Chr.1: 215586685-215586685 on Build GRCh38
rs765208259 Chr.1: 215586708-215586709 on Build GRCh38
rs778771433 Chr.1: 215586712-215586712 on Build GRCh38
rs757857647 Chr.1: 215586724-215586724 on Build GRCh38
rs766187565 Chr.1: 215586706-215586706 on Build GRCh38
rs749205838 Chr.1: 215586556-215586556 on Build GRCh38
rs145226766 Chr.1: 215586638-215586638 on Build GRCh38
rs754885361 Chr.1: 215586711-215586711 on Build GRCh38
rs372159813 Chr.1: 215586666-215586666 on Build GRCh38
rs375582499 Chr.1: 215586611-215586611 on Build GRCh38
rs763551206 Chr.1: 215586646-215586646 on Build GRCh38

View Full Product Details