Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26426371-26426610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIN28A
Entrez Gene ID
79727
Gene Name
lin-28 homolog A
Gene Aliases
CSDD1, LIN-28, LIN28, ZCCHC1, lin-28A
UniGene
Hs.86154
Gene Chromosome Location
Chr.1: 26410768-26429728 on Build GRCh38
Transcripts
NM_024674.5 XM_011542148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5533824 c.557G>A p.G186E Chr.1: 26426385-26426385 on Build GRCh38
5575122 c.610C>T p.L204F Chr.1: 26426438-26426438 on Build GRCh38
1685149 c.615G>A p.P205P Chr.1: 26426443-26426443 on Build GRCh38
3671704 c.614C>A p.P205Q Chr.1: 26426442-26426442 on Build GRCh38
7253075 c.545C>A p.P182H Chr.1: 26426373-26426373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770384641 Chr.1: 26426476-26426476 on Build GRCh38
rs765330629 Chr.1: 26426432-26426432 on Build GRCh38
rs199541048 Chr.1: 26426465-26426465 on Build GRCh38
rs377421420 Chr.1: 26426396-26426396 on Build GRCh38
rs201055562 Chr.1: 26426443-26426443 on Build GRCh38
rs558060339 Chr.1: 26426402-26426402 on Build GRCh38
rs919714326 Chr.1: 26426586-26426586 on Build GRCh38
rs928365608 Chr.1: 26426541-26426541 on Build GRCh38
rs757331207 Chr.1: 26426428-26426428 on Build GRCh38
rs998972819 Chr.1: 26426460-26426460 on Build GRCh38
rs771859394 Chr.1: 26426504-26426504 on Build GRCh38
rs201124162 Chr.1: 26426394-26426394 on Build GRCh38
rs748777822 Chr.1: 26426391-26426391 on Build GRCh38
rs777591401 Chr.1: 26426475-26426475 on Build GRCh38
rs758605588 Chr.1: 26426440-26426440 on Build GRCh38
rs377434392 Chr.1: 26426468-26426468 on Build GRCh38
rs972568456 Chr.1: 26426578-26426578 on Build GRCh38
rs764543175 Chr.1: 26426405-26426405 on Build GRCh38
rs769814831 Chr.1: 26426533-26426533 on Build GRCh38
rs768055786 Chr.1: 26426397-26426397 on Build GRCh38
rs970116417 Chr.1: 26426525-26426525 on Build GRCh38
rs370187333 Chr.1: 26426433-26426433 on Build GRCh38
rs576059503 Chr.1: 26426566-26426566 on Build GRCh38
rs748187241 Chr.1: 26426403-26426403 on Build GRCh38
rs781122495 Chr.1: 26426408-26426410 on Build GRCh38
rs754323496 Chr.1: 26426407-26426407 on Build GRCh38
rs939868666 Chr.1: 26426576-26426577 on Build GRCh38
rs747220883 Chr.1: 26426502-26426502 on Build GRCh38
rs556708356 Chr.1: 26426585-26426585 on Build GRCh38
rs775246055 Chr.1: 26426505-26426505 on Build GRCh38
rs4659441 Chr.1: 26426501-26426501 on Build GRCh38

View Full Product Details