Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48334324-48334597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUT
Entrez Gene ID
1854
Gene Name
deoxyuridine triphosphatase
Gene Aliases
dUTPase
UniGene
Hs.527980
Gene Chromosome Location
Chr.15: 48331018-48343373 on Build GRCh38
Transcripts
NM_001025248.1 NM_001025249.1 NM_001948.3 XM_011521315.2 XM_017021988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4055294 c.167A>G p.Y56C Chr.15: 48334428-48334428 on Build GRCh38
5458502 c.482C>T p.A161V Chr.15: 48334479-48334479 on Build GRCh38
6650424 c.180T>C p.P60P Chr.15: 48334441-48334441 on Build GRCh38
2190245 c.229delG p.C78fs*19 Chr.15: 48334490-48334490 on Build GRCh38
2190244 c.493delG p.C166fs*19 Chr.15: 48334490-48334490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201357294 Chr.15: 48334480-48334480 on Build GRCh38
rs770412730 Chr.15: 48334532-48334532 on Build GRCh38
rs762653430 Chr.15: 48334438-48334438 on Build GRCh38
rs868462783 Chr.15: 48334540-48334540 on Build GRCh38
rs757373173 Chr.15: 48334525-48334525 on Build GRCh38
rs201667550 Chr.15: 48334414-48334414 on Build GRCh38
rs751398863 Chr.15: 48334479-48334479 on Build GRCh38
rs777840231 Chr.15: 48334376-48334376 on Build GRCh38
rs777786832 Chr.15: 48334498-48334498 on Build GRCh38
rs776311907 Chr.15: 48334413-48334413 on Build GRCh38
rs772543234 Chr.15: 48334394-48334394 on Build GRCh38
rs746194583 Chr.15: 48334388-48334388 on Build GRCh38
rs565876267 Chr.15: 48334529-48334529 on Build GRCh38
rs28381112 Chr.15: 48334450-48334450 on Build GRCh38
rs750160491 Chr.15: 48334425-48334425 on Build GRCh38
rs1005448613 Chr.15: 48334412-48334412 on Build GRCh38
rs780789372 Chr.15: 48334385-48334385 on Build GRCh38
rs373184762 Chr.15: 48334422-48334422 on Build GRCh38
rs753882201 Chr.15: 48334500-48334500 on Build GRCh38
rs779092209 Chr.15: 48334527-48334527 on Build GRCh38
rs755436547 Chr.15: 48334353-48334356 on Build GRCh38
rs561345005 Chr.15: 48334371-48334371 on Build GRCh38
rs185362945 Chr.15: 48334365-48334365 on Build GRCh38
rs774594062 Chr.15: 48334381-48334381 on Build GRCh38
rs1047322990 Chr.15: 48334404-48334404 on Build GRCh38
rs775856887 Chr.15: 48334397-48334397 on Build GRCh38
rs888340233 Chr.15: 48334407-48334407 on Build GRCh38
rs200190942 Chr.15: 48334399-48334399 on Build GRCh38
rs201190039 Chr.15: 48334437-48334437 on Build GRCh38
rs368212041 Chr.15: 48334377-48334377 on Build GRCh38
rs1055496942 Chr.15: 48334469-48334469 on Build GRCh38
rs190555788 Chr.15: 48334405-48334405 on Build GRCh38
rs776380602 Chr.15: 48334440-48334440 on Build GRCh38
rs771171217 Chr.15: 48334378-48334378 on Build GRCh38

View Full Product Details