Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101827548-101827791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNIP2
Entrez Gene ID
30819
Gene Name
potassium voltage-gated channel interacting protein 2
Gene Aliases
KCHIP2
UniGene
Hs.97044
Gene Chromosome Location
Chr.10: 101825971-101843920 on Build GRCh38
Transcripts
NM_014591.4 NM_173191.2 NM_173192.2 NM_173193.2 NM_173194.2 NM_173195.2 NM_173197.2 XM_005269729.2 XM_005269730.2 XM_006717812.2 XM_011539731.2 XM_017016161.1 NR_045118.1

Gene Symbol
KCNIP2-AS1
Entrez Gene ID
100289509
Gene Name
KCNIP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.568881
Gene Chromosome Location
Chr.10: 101819068-101828889 on Build GRCh38
Transcripts
NM_014591.4 NM_173191.2 NM_173192.2 NM_173193.2 NM_173194.2 NM_173195.2 NM_173197.2 XM_005269729.2 XM_005269730.2 XM_006717812.2 XM_011539731.2 XM_017016161.1 NR_045118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997956 c.528C>G p.Y176* Chr.10: 101827668-101827668 on Build GRCh38
4833380 c.554G>C p.*185S Chr.10: 101827642-101827642 on Build GRCh38
7147480 c.525C>G p.L175L Chr.10: 101827671-101827671 on Build GRCh38
4165374 c.624T>C p.C208C Chr.10: 101827695-101827695 on Build GRCh38
4165373 c.804T>C p.C268C Chr.10: 101827695-101827695 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778052779 Chr.10: 101827618-101827618 on Build GRCh38
rs773791083 Chr.10: 101827657-101827657 on Build GRCh38
rs759484365 Chr.10: 101827707-101827707 on Build GRCh38
rs951526453 Chr.10: 101827749-101827749 on Build GRCh38
rs907144743 Chr.10: 101827569-101827569 on Build GRCh38
rs752768127 Chr.10: 101827596-101827596 on Build GRCh38
rs1034058019 Chr.10: 101827626-101827626 on Build GRCh38
rs776347807 Chr.10: 101827705-101827705 on Build GRCh38
rs764275518 Chr.10: 101827759-101827759 on Build GRCh38
rs752790110 Chr.10: 101827721-101827721 on Build GRCh38
rs772896557 Chr.10: 101827679-101827679 on Build GRCh38
rs751859470 Chr.10: 101827769-101827769 on Build GRCh38
rs760269458 Chr.10: 101827682-101827682 on Build GRCh38
rs368114335 Chr.10: 101827603-101827603 on Build GRCh38
rs774985846 Chr.10: 101827749-101827749 on Build GRCh38
rs992336586 Chr.10: 101827728-101827728 on Build GRCh38
rs769441984 Chr.10: 101827661-101827661 on Build GRCh38
rs747375622 Chr.10: 101827632-101827632 on Build GRCh38
rs747685150 Chr.10: 101827662-101827662 on Build GRCh38
rs771658331 Chr.10: 101827670-101827670 on Build GRCh38
rs374874936 Chr.10: 101827717-101827717 on Build GRCh38
rs781654565 Chr.10: 101827646-101827646 on Build GRCh38
rs758477233 Chr.10: 101827737-101827737 on Build GRCh38
rs766195922 Chr.10: 101827694-101827694 on Build GRCh38
rs536763907 Chr.10: 101827642-101827642 on Build GRCh38
rs1001597455 Chr.10: 101827584-101827584 on Build GRCh38
rs371574418 Chr.10: 101827649-101827649 on Build GRCh38

View Full Product Details