Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98128550-98128789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO2A
Entrez Gene ID
7464
Gene Name
coronin 2A
Gene Aliases
CLIPINB, IR10, WDR2
UniGene
Hs.113094
Gene Chromosome Location
Chr.9: 98120975-98192674 on Build GRCh38
Transcripts
NM_003389.3 NM_052820.3 XM_011518986.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7095264 c.1065G>A p.M355I Chr.9: 98128622-98128622 on Build GRCh38
2793305 c.1050C>T p.I350I Chr.9: 98128637-98128637 on Build GRCh38
2793306 c.990C>T p.L330L Chr.9: 98128697-98128697 on Build GRCh38
7137383 c.1047C>T p.L349L Chr.9: 98128640-98128640 on Build GRCh38
3652196 c.1056C>T p.P352P Chr.9: 98128631-98128631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370084835 Chr.9: 98128594-98128594 on Build GRCh38
rs745399135 Chr.9: 98128630-98128630 on Build GRCh38
rs141466107 Chr.9: 98128609-98128609 on Build GRCh38
rs754676609 Chr.9: 98128769-98128769 on Build GRCh38
rs771869570 Chr.9: 98128582-98128582 on Build GRCh38
rs775753856 Chr.9: 98128686-98128686 on Build GRCh38
rs375771898 Chr.9: 98128589-98128589 on Build GRCh38
rs73655194 Chr.9: 98128672-98128672 on Build GRCh38
rs1051889187 Chr.9: 98128615-98128615 on Build GRCh38
rs776493594 Chr.9: 98128670-98128670 on Build GRCh38
rs762181812 Chr.9: 98128719-98128719 on Build GRCh38
rs991541258 Chr.9: 98128588-98128588 on Build GRCh38
rs368276546 Chr.9: 98128697-98128697 on Build GRCh38
rs772266826 Chr.9: 98128681-98128681 on Build GRCh38
rs749929107 Chr.9: 98128734-98128734 on Build GRCh38
rs754231931 Chr.9: 98128614-98128614 on Build GRCh38
rs765882359 Chr.9: 98128733-98128733 on Build GRCh38
rs377513173 Chr.9: 98128747-98128747 on Build GRCh38
rs760955757 Chr.9: 98128693-98128693 on Build GRCh38
rs778513387 Chr.9: 98128624-98128624 on Build GRCh38
rs779898610 Chr.9: 98128636-98128636 on Build GRCh38
rs749492230 Chr.9: 98128737-98128737 on Build GRCh38
rs368531711 Chr.9: 98128662-98128662 on Build GRCh38
rs775383090 Chr.9: 98128586-98128586 on Build GRCh38
rs780280366 Chr.9: 98128684-98128686 on Build GRCh38
rs764655582 Chr.9: 98128612-98128612 on Build GRCh38
rs1035406194 Chr.9: 98128761-98128761 on Build GRCh38
rs928761823 Chr.9: 98128576-98128576 on Build GRCh38
rs375182845 Chr.9: 98128692-98128692 on Build GRCh38
rs747978763 Chr.9: 98128671-98128671 on Build GRCh38
rs373619389 Chr.9: 98128600-98128600 on Build GRCh38
rs571776662 Chr.9: 98128634-98128634 on Build GRCh38
rs865850359 Chr.9: 98128575-98128575 on Build GRCh38
rs147095167 Chr.9: 98128658-98128658 on Build GRCh38
rs756492505 Chr.9: 98128611-98128611 on Build GRCh38
rs746789995 Chr.9: 98128637-98128637 on Build GRCh38
rs759654091 Chr.9: 98128679-98128679 on Build GRCh38
rs757796002 Chr.9: 98128618-98128618 on Build GRCh38
rs758042497 Chr.9: 98128742-98128742 on Build GRCh38
rs768618360 Chr.9: 98128593-98128593 on Build GRCh38
rs367766736 Chr.9: 98128595-98128595 on Build GRCh38
rs894065931 Chr.9: 98128682-98128682 on Build GRCh38
rs923787308 Chr.9: 98128642-98128642 on Build GRCh38
rs779851447 Chr.9: 98128743-98128743 on Build GRCh38
rs767760132 Chr.9: 98128608-98128608 on Build GRCh38
rs768406146 Chr.9: 98128667-98128667 on Build GRCh38
rs759707070 Chr.9: 98128601-98128601 on Build GRCh38

View Full Product Details