Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48302809-48303074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC114
Entrez Gene ID
93233
Gene Name
coiled-coil domain containing 114
Gene Aliases
CILD20
UniGene
Hs.112645
Gene Chromosome Location
Chr.19: 48296452-48322793 on Build GRCh38
Transcripts
NM_144577.3 XM_005259413.3 XM_005259414.3 XM_005259415.3 XM_005259416.4 XM_011527515.2 XM_011527516.2 XM_011527517.1 XM_017027483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7120350 c.325G>A p.E109K Chr.19: 48303027-48303027 on Build GRCh38
4079825 c.985C>T p.R329C Chr.19: 48302838-48302838 on Build GRCh38
384922 c.982G>A p.A328T Chr.19: 48302841-48302841 on Build GRCh38
243513 c.915C>T p.N305N Chr.19: 48303058-48303058 on Build GRCh38
191491 c.364C>T p.R122C Chr.19: 48302838-48302838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753922343 Chr.19: 48302861-48302861 on Build GRCh38
rs561780439 Chr.19: 48302841-48302841 on Build GRCh38
rs751571535 Chr.19: 48302842-48302842 on Build GRCh38
rs774088366 Chr.19: 48303049-48303049 on Build GRCh38
rs748648447 Chr.19: 48303018-48303018 on Build GRCh38
rs747629232 Chr.19: 48302909-48302909 on Build GRCh38
rs606231240 Chr.19: 48303034-48303034 on Build GRCh38
rs532136141 Chr.19: 48302904-48302904 on Build GRCh38
rs548341766 Chr.19: 48302989-48302989 on Build GRCh38
rs530604105 Chr.19: 48302896-48302896 on Build GRCh38
rs767556426 Chr.19: 48302857-48302857 on Build GRCh38
rs374566612 Chr.19: 48302903-48302903 on Build GRCh38
rs370741101 Chr.19: 48302969-48302969 on Build GRCh38
rs370945527 Chr.19: 48302882-48302882 on Build GRCh38
rs372553613 Chr.19: 48302831-48302831 on Build GRCh38
rs370315745 Chr.19: 48302913-48302913 on Build GRCh38
rs368014089 Chr.19: 48302984-48302984 on Build GRCh38
rs745662455 Chr.19: 48303050-48303050 on Build GRCh38
rs769393527 Chr.19: 48302838-48302838 on Build GRCh38
rs758656819 Chr.19: 48302986-48302986 on Build GRCh38
rs772867955 Chr.19: 48302839-48302839 on Build GRCh38
rs946093311 Chr.19: 48302972-48302972 on Build GRCh38
rs938203086 Chr.19: 48302975-48302975 on Build GRCh38
rs534910262 Chr.19: 48302981-48302981 on Build GRCh38
rs758781406 Chr.19: 48302894-48302894 on Build GRCh38
rs1043921424 Chr.19: 48302964-48302964 on Build GRCh38
rs1016032486 Chr.19: 48302859-48302859 on Build GRCh38
rs375985392 Chr.19: 48302911-48302911 on Build GRCh38
rs566316291 Chr.19: 48302949-48302949 on Build GRCh38
rs922700741 Chr.19: 48302854-48302854 on Build GRCh38
rs755602988 Chr.19: 48303019-48303019 on Build GRCh38
rs777393933 Chr.19: 48303029-48303029 on Build GRCh38
rs774352752 Chr.19: 48302962-48302962 on Build GRCh38
rs58694009 Chr.19: 48302877-48302877 on Build GRCh38
rs375357239 Chr.19: 48302973-48302973 on Build GRCh38
rs79967289 Chr.19: 48303013-48303013 on Build GRCh38
rs747346139 Chr.19: 48302897-48302897 on Build GRCh38
rs35361179 Chr.19: 48302837-48302837 on Build GRCh38
rs759585116 Chr.19: 48302855-48302855 on Build GRCh38
rs1029994161 Chr.19: 48302840-48302840 on Build GRCh38
rs755281213 Chr.19: 48303003-48303003 on Build GRCh38
rs190428141 Chr.19: 48302915-48302915 on Build GRCh38
rs200966511 Chr.19: 48303031-48303031 on Build GRCh38
rs376126041 Chr.19: 48302963-48302963 on Build GRCh38
rs779002861 Chr.19: 48302870-48302870 on Build GRCh38
rs757303254 Chr.19: 48302863-48302863 on Build GRCh38
rs770763751 Chr.19: 48303046-48303046 on Build GRCh38
rs563921813 Chr.19: 48302912-48302912 on Build GRCh38
rs116228677 Chr.19: 48302990-48302990 on Build GRCh38
rs774616177 Chr.19: 48302985-48302985 on Build GRCh38

View Full Product Details