Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32310384-32310655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF3B
Entrez Gene ID
9371
Gene Name
kinesin family member 3B
Gene Aliases
FLA8, HH0048, KLP-11
UniGene
Hs.369670
Gene Chromosome Location
Chr.20: 32277640-32335011 on Build GRCh38
Transcripts
NM_004798.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5883541 c.812C>T p.S271F Chr.20: 32310589-32310589 on Build GRCh38
1196791 c.875A>G p.Y292C Chr.20: 32310652-32310652 on Build GRCh38
6695503 c.690C>T p.L230L Chr.20: 32310467-32310467 on Build GRCh38
6695505 c.696T>G p.G232G Chr.20: 32310473-32310473 on Build GRCh38
4822355 c.844C>G p.L282V Chr.20: 32310621-32310621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940799632 Chr.20: 32310597-32310597 on Build GRCh38
rs781040842 Chr.20: 32310461-32310461 on Build GRCh38
rs755570657 Chr.20: 32310472-32310472 on Build GRCh38
rs763920518 Chr.20: 32310451-32310451 on Build GRCh38
rs773609367 Chr.20: 32310536-32310536 on Build GRCh38
rs770139100 Chr.20: 32310502-32310502 on Build GRCh38
rs916976056 Chr.20: 32310631-32310631 on Build GRCh38
rs767463571 Chr.20: 32310617-32310617 on Build GRCh38
rs1033380046 Chr.20: 32310410-32310410 on Build GRCh38
rs1054649511 Chr.20: 32310435-32310435 on Build GRCh38
rs745395716 Chr.20: 32310467-32310467 on Build GRCh38
rs763217315 Chr.20: 32310569-32310569 on Build GRCh38
rs751501042 Chr.20: 32310455-32310455 on Build GRCh38
rs774113271 Chr.20: 32310611-32310611 on Build GRCh38
rs1018945730 Chr.20: 32310468-32310468 on Build GRCh38
rs138618271 Chr.20: 32310596-32310596 on Build GRCh38
rs937022378 Chr.20: 32310420-32310420 on Build GRCh38
rs370499804 Chr.20: 32310418-32310418 on Build GRCh38
rs200496440 Chr.20: 32310545-32310545 on Build GRCh38
rs926946845 Chr.20: 32310416-32310416 on Build GRCh38
rs202021047 Chr.20: 32310524-32310524 on Build GRCh38
rs866834295 Chr.20: 32310440-32310440 on Build GRCh38
rs138197776 Chr.20: 32310456-32310456 on Build GRCh38
rs756943920 Chr.20: 32310614-32310614 on Build GRCh38
rs779710160 Chr.20: 32310473-32310473 on Build GRCh38
rs562693298 Chr.20: 32310560-32310560 on Build GRCh38
rs146174510 Chr.20: 32310542-32310542 on Build GRCh38
rs755658567 Chr.20: 32310630-32310630 on Build GRCh38
rs149299393 Chr.20: 32310629-32310629 on Build GRCh38
rs745618066 Chr.20: 32310423-32310424 on Build GRCh38
rs200794805 Chr.20: 32310491-32310491 on Build GRCh38
rs368533621 Chr.20: 32310563-32310563 on Build GRCh38
rs771476770 Chr.20: 32310543-32310543 on Build GRCh38
rs759713888 Chr.20: 32310546-32310546 on Build GRCh38
rs142727710 Chr.20: 32310487-32310487 on Build GRCh38
rs758159723 Chr.20: 32310450-32310450 on Build GRCh38
rs765572997 Chr.20: 32310404-32310404 on Build GRCh38
rs966117347 Chr.20: 32310501-32310501 on Build GRCh38

View Full Product Details