Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30370672-30370917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC157
Entrez Gene ID
550631
Gene Name
coiled-coil domain containing 157
Gene Aliases
-
UniGene
Hs.505597
Gene Chromosome Location
Chr.22: 30356635-30378655 on Build GRCh38
Transcripts
NM_001017437.3 NM_001318334.1 XM_006724278.3 XM_006724279.3 XM_006724280.3 XM_011530263.2 XM_011530266.2 XM_011530267.2 XM_011530268.2 XM_011530270.2 NR_046312.1

Gene Symbol
KIAA1656
Entrez Gene ID
85371
Gene Name
KIAA1656 protein
Gene Aliases
-
UniGene
Hs.505597
Gene Chromosome Location
Chr.22: 30368811-30377905 on Build GRCh38
Transcripts
NM_001017437.3 NM_001318334.1 XM_006724278.3 XM_006724279.3 XM_006724280.3 XM_011530263.2 XM_011530266.2 XM_011530267.2 XM_011530268.2 XM_011530270.2 NR_046312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3379296 c.884C>T p.A295V Chr.22: 30370789-30370789 on Build GRCh38
6621766 c.986C>T p.A329V Chr.22: 30370891-30370891 on Build GRCh38
5687899 c.911A>G p.K304R Chr.22: 30370816-30370816 on Build GRCh38
6621764 c.973C>T p.R325W Chr.22: 30370878-30370878 on Build GRCh38
5687900 c.758A>G p.K253R Chr.22: 30370816-30370816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752251966 Chr.22: 30370715-30370715 on Build GRCh38
rs377404857 Chr.22: 30370845-30370845 on Build GRCh38
rs546033949 Chr.22: 30370855-30370855 on Build GRCh38
rs547209278 Chr.22: 30370803-30370803 on Build GRCh38
rs141347000 Chr.22: 30370892-30370892 on Build GRCh38
rs763533762 Chr.22: 30370797-30370797 on Build GRCh38
rs1004947996 Chr.22: 30370723-30370723 on Build GRCh38
rs759472730 Chr.22: 30370772-30370772 on Build GRCh38
rs772145253 Chr.22: 30370837-30370837 on Build GRCh38
rs780962836 Chr.22: 30370818-30370818 on Build GRCh38
rs751163184 Chr.22: 30370801-30370801 on Build GRCh38
rs746672484 Chr.22: 30370726-30370726 on Build GRCh38
rs755387549 Chr.22: 30370828-30370828 on Build GRCh38
rs541801601 Chr.22: 30370776-30370776 on Build GRCh38
rs760492483 Chr.22: 30370692-30370692 on Build GRCh38
rs143230422 Chr.22: 30370861-30370861 on Build GRCh38
rs775520038 Chr.22: 30370857-30370857 on Build GRCh38
rs141091935 Chr.22: 30370709-30370709 on Build GRCh38
rs200598609 Chr.22: 30370769-30370769 on Build GRCh38
rs766122756 Chr.22: 30370693-30370693 on Build GRCh38
rs553097316 Chr.22: 30370728-30370728 on Build GRCh38
rs761441332 Chr.22: 30370865-30370865 on Build GRCh38
rs1024004607 Chr.22: 30370891-30370891 on Build GRCh38
rs766590165 Chr.22: 30370864-30370865 on Build GRCh38
rs143878154 Chr.22: 30370838-30370838 on Build GRCh38
rs760833855 Chr.22: 30370760-30370760 on Build GRCh38
rs762811184 Chr.22: 30370858-30370858 on Build GRCh38
rs138237270 Chr.22: 30370768-30370768 on Build GRCh38
rs148283823 Chr.22: 30370879-30370879 on Build GRCh38
rs763590195 Chr.22: 30370859-30370859 on Build GRCh38
rs374269153 Chr.22: 30370814-30370814 on Build GRCh38
rs753761288 Chr.22: 30370698-30370698 on Build GRCh38
rs757942184 Chr.22: 30370790-30370790 on Build GRCh38
rs867844733 Chr.22: 30370812-30370812 on Build GRCh38
rs774892887 Chr.22: 30370746-30370746 on Build GRCh38
rs765243365 Chr.22: 30370775-30370775 on Build GRCh38
rs765153541 Chr.22: 30370856-30370856 on Build GRCh38
rs758030115 Chr.22: 30370718-30370718 on Build GRCh38
rs546676831 Chr.22: 30370885-30370885 on Build GRCh38
rs754979631 Chr.22: 30370708-30370708 on Build GRCh38
rs750119069 Chr.22: 30370882-30370882 on Build GRCh38
rs749724460 Chr.22: 30370737-30370737 on Build GRCh38
rs748207336 Chr.22: 30370756-30370756 on Build GRCh38
rs149837002 Chr.22: 30370719-30370719 on Build GRCh38
rs755299220 Chr.22: 30370884-30370884 on Build GRCh38
rs145788485 Chr.22: 30370730-30370730 on Build GRCh38
rs563598346 Chr.22: 30370831-30370831 on Build GRCh38
rs747188480 Chr.22: 30370839-30370839 on Build GRCh38
rs758834156 Chr.22: 30370898-30370898 on Build GRCh38
rs879034884 Chr.22: 30370725-30370725 on Build GRCh38
rs749636612 Chr.22: 30370825-30370825 on Build GRCh38
rs145673028 Chr.22: 30370758-30370758 on Build GRCh38
rs772087168 Chr.22: 30370757-30370757 on Build GRCh38
rs748689658 Chr.22: 30370836-30370836 on Build GRCh38
rs942927542 Chr.22: 30370759-30370759 on Build GRCh38
rs769144873 Chr.22: 30370744-30370744 on Build GRCh38
rs779364454 Chr.22: 30370830-30370830 on Build GRCh38

View Full Product Details