Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23798786-23799043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALE
Entrez Gene ID
2582
Gene Name
UDP-galactose-4-epimerase
Gene Aliases
SDR1E1
UniGene
Hs.632380
Gene Chromosome Location
Chr.1: 23795599-23800804 on Build GRCh38
Transcripts
NM_000403.3 NM_001008216.1 NM_001127621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1243279 c.95T>G p.I32S Chr.1: 23798913-23798913 on Build GRCh38
1626930 c.1A>G p.M1V Chr.1: 23799007-23799007 on Build GRCh38
6062522 c.105C>G p.F35L Chr.1: 23798903-23798903 on Build GRCh38
3400552 c.7G>C p.E3Q Chr.1: 23799001-23799001 on Build GRCh38
4612660 c.23_24insC p.G9fs*26 Chr.1: 23798984-23798985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969033368 Chr.1: 23798812-23798812 on Build GRCh38
rs754537742 Chr.1: 23798840-23798840 on Build GRCh38
rs747590712 Chr.1: 23798894-23798894 on Build GRCh38
rs766676772 Chr.1: 23798918-23798918 on Build GRCh38
rs777541696 Chr.1: 23798939-23798939 on Build GRCh38
rs766058013 Chr.1: 23798808-23798808 on Build GRCh38
rs754850112 Chr.1: 23798977-23798977 on Build GRCh38
rs375172302 Chr.1: 23798854-23798854 on Build GRCh38
rs140540126 Chr.1: 23798813-23798813 on Build GRCh38
rs772953121 Chr.1: 23798897-23798897 on Build GRCh38
rs762599048 Chr.1: 23798917-23798917 on Build GRCh38
rs748115409 Chr.1: 23798989-23798989 on Build GRCh38
rs1047916743 Chr.1: 23798895-23798895 on Build GRCh38
rs121908046 Chr.1: 23798907-23798907 on Build GRCh38
rs890475979 Chr.1: 23798821-23798821 on Build GRCh38
rs758390003 Chr.1: 23798921-23798921 on Build GRCh38
rs746828892 Chr.1: 23798955-23798955 on Build GRCh38
rs794727483 Chr.1: 23798959-23798959 on Build GRCh38
rs150919841 Chr.1: 23798931-23798931 on Build GRCh38
rs764270555 Chr.1: 23798944-23798944 on Build GRCh38
rs200873266 Chr.1: 23798881-23798881 on Build GRCh38
rs144492228 Chr.1: 23798890-23798890 on Build GRCh38
rs779261698 Chr.1: 23798973-23798973 on Build GRCh38
rs532648624 Chr.1: 23798928-23798928 on Build GRCh38
rs28719646 Chr.1: 23798832-23798832 on Build GRCh38
rs772725720 Chr.1: 23799010-23799010 on Build GRCh38
rs756309437 Chr.1: 23798847-23798847 on Build GRCh38
rs753266922 Chr.1: 23798941-23798941 on Build GRCh38
rs762637068 Chr.1: 23798907-23798908 on Build GRCh38
rs727503945 Chr.1: 23798911-23798911 on Build GRCh38
rs756219818 Chr.1: 23798942-23798942 on Build GRCh38
rs755457329 Chr.1: 23798836-23798836 on Build GRCh38
rs769561998 Chr.1: 23798875-23798875 on Build GRCh38
rs776668071 Chr.1: 23798893-23798893 on Build GRCh38
rs796065329 Chr.1: 23798911-23798913 on Build GRCh38
rs758195864 Chr.1: 23798869-23798869 on Build GRCh38
rs780943903 Chr.1: 23798980-23798980 on Build GRCh38
rs900472958 Chr.1: 23798999-23798999 on Build GRCh38
rs769318489 Chr.1: 23798902-23798902 on Build GRCh38
rs752718155 Chr.1: 23798845-23798845 on Build GRCh38
rs769226903 Chr.1: 23799008-23799008 on Build GRCh38
rs904759055 Chr.1: 23798880-23798880 on Build GRCh38
rs778435909 Chr.1: 23798889-23798889 on Build GRCh38
rs770785773 Chr.1: 23798947-23798988 on Build GRCh38
rs772813271 Chr.1: 23798912-23798912 on Build GRCh38
rs746748470 Chr.1: 23798976-23798976 on Build GRCh38
rs373419146 Chr.1: 23798852-23798852 on Build GRCh38
rs774570531 Chr.1: 23798923-23798923 on Build GRCh38
rs143264410 Chr.1: 23798967-23798967 on Build GRCh38
rs187654497 Chr.1: 23798954-23798954 on Build GRCh38

View Full Product Details