Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 132632753-132633014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC6
Entrez Gene ID
23639
Gene Name
leucine rich repeat containing 6
Gene Aliases
CILD19, LRTP, TSLRP
UniGene
Hs.591865
Gene Chromosome Location
Chr.8: 132570419-132685039 on Build GRCh38
Transcripts
NM_001321961.1 NM_001321962.1 NM_001321963.1 NM_001321964.1 NM_001321965.1 NM_001321966.1 NM_012472.5 NR_073525.2 NR_135905.1 NR_135906.1 NR_135907.1 NR_135908.1 NR_135909.1 NR_135910.1 NR_135911.1 NR_135912.1 NR_135913.1 XM_006716538.3 XM_011516950.2 XM_017013296.1 XM_017013297.1 XM_017013298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7375646 c.574C>G p.Q192E Chr.8: 132632819-132632819 on Build GRCh38
5534039 c.464G>A p.R155K Chr.8: 132632929-132632929 on Build GRCh38
5552931 c.518A>G p.E173G Chr.8: 132632875-132632875 on Build GRCh38
1096236 c.457T>G p.S153A Chr.8: 132632936-132632936 on Build GRCh38
1096237 c.445G>T p.E149* Chr.8: 132632948-132632948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376633613 Chr.8: 132632782-132632782 on Build GRCh38
rs536226289 Chr.8: 132632884-132632884 on Build GRCh38
rs758765614 Chr.8: 132632817-132632817 on Build GRCh38
rs941424900 Chr.8: 132632840-132632840 on Build GRCh38
rs766487177 Chr.8: 132632885-132632885 on Build GRCh38
rs1030772313 Chr.8: 132632775-132632775 on Build GRCh38
rs146136398 Chr.8: 132632921-132632921 on Build GRCh38
rs754431068 Chr.8: 132632968-132632968 on Build GRCh38
rs779571778 Chr.8: 132632948-132632948 on Build GRCh38
rs751836811 Chr.8: 132632939-132632939 on Build GRCh38
rs753177783 Chr.8: 132632797-132632797 on Build GRCh38
rs772453135 Chr.8: 132632783-132632783 on Build GRCh38
rs753194043 Chr.8: 132632904-132632904 on Build GRCh38
rs200321595 Chr.8: 132632957-132632957 on Build GRCh38
rs748037780 Chr.8: 132632972-132632972 on Build GRCh38
rs1051351752 Chr.8: 132632982-132632982 on Build GRCh38
rs1056908657 Chr.8: 132632912-132632912 on Build GRCh38
rs755856484 Chr.8: 132632941-132632941 on Build GRCh38
rs768652184 Chr.8: 132632849-132632849 on Build GRCh38
rs397515424 Chr.8: 132632794-132632795 on Build GRCh38
rs397515425 Chr.8: 132632816-132632817 on Build GRCh38
rs1037263244 Chr.8: 132632979-132632979 on Build GRCh38
rs369802490 Chr.8: 132632879-132632879 on Build GRCh38
rs367605857 Chr.8: 132632862-132632862 on Build GRCh38
rs141945265 Chr.8: 132632819-132632819 on Build GRCh38
rs748737174 Chr.8: 132632843-132632843 on Build GRCh38
rs759683541 Chr.8: 132632889-132632889 on Build GRCh38
rs774210701 Chr.8: 132632854-132632854 on Build GRCh38
rs371789072 Chr.8: 132632867-132632867 on Build GRCh38
rs779550260 Chr.8: 132632792-132632792 on Build GRCh38
rs938070612 Chr.8: 132632880-132632880 on Build GRCh38
rs762626179 Chr.8: 132632880-132632881 on Build GRCh38
rs771806569 Chr.8: 132632865-132632865 on Build GRCh38
rs35956157 Chr.8: 132632784-132632784 on Build GRCh38
rs776827950 Chr.8: 132632886-132632886 on Build GRCh38
rs1038439690 Chr.8: 132632892-132632892 on Build GRCh38
rs778844790 Chr.8: 132632970-132632970 on Build GRCh38
rs1003794367 Chr.8: 132632798-132632798 on Build GRCh38
rs769215386 Chr.8: 132632828-132632828 on Build GRCh38
rs397515461 Chr.8: 132632831-132632831 on Build GRCh38
rs769547620 Chr.8: 132632847-132632847 on Build GRCh38
rs765843715 Chr.8: 132632903-132632903 on Build GRCh38
rs568785915 Chr.8: 132632787-132632787 on Build GRCh38
rs979934112 Chr.8: 132632855-132632855 on Build GRCh38

View Full Product Details