Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53213355-53213628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT2
Entrez Gene ID
1376
Gene Name
carnitine palmitoyltransferase 2
Gene Aliases
CPT1, CPTASE, IIAE4
UniGene
Hs.713535
Gene Chromosome Location
Chr.1: 53196429-53214197 on Build GRCh38
Transcripts
NM_000098.2 XM_005270484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202215 c.1751A>C p.H584P Chr.1: 53213369-53213369 on Build GRCh38
6634987 c.1881C>A p.S627S Chr.1: 53213499-53213499 on Build GRCh38
5482421 c.1891C>T p.R631C Chr.1: 53213509-53213509 on Build GRCh38
7166513 c.1766C>T p.T589M Chr.1: 53213384-53213384 on Build GRCh38
6634982 c.1756G>T p.V586F Chr.1: 53213374-53213374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769539793 Chr.1: 53213412-53213412 on Build GRCh38
rs1042375038 Chr.1: 53213512-53213512 on Build GRCh38
rs147953465 Chr.1: 53213424-53213424 on Build GRCh38
rs761631943 Chr.1: 53213496-53213496 on Build GRCh38
rs515726179 Chr.1: 53213541-53213553 on Build GRCh38
rs1799822 Chr.1: 53213557-53213557 on Build GRCh38
rs901767832 Chr.1: 53213502-53213502 on Build GRCh38
rs752874182 Chr.1: 53213520-53213520 on Build GRCh38
rs767530116 Chr.1: 53213504-53213504 on Build GRCh38
rs758803512 Chr.1: 53213531-53213531 on Build GRCh38
rs748946176 Chr.1: 53213555-53213555 on Build GRCh38
rs769108011 Chr.1: 53213390-53213390 on Build GRCh38
rs1009503062 Chr.1: 53213510-53213510 on Build GRCh38
rs201913567 Chr.1: 53213438-53213438 on Build GRCh38
rs373714948 Chr.1: 53213582-53213582 on Build GRCh38
rs759101149 Chr.1: 53213409-53213409 on Build GRCh38
rs796051943 Chr.1: 53213407-53213407 on Build GRCh38
rs201996784 Chr.1: 53213467-53213467 on Build GRCh38
rs1871748 Chr.1: 53213381-53213381 on Build GRCh38
rs778379750 Chr.1: 53213542-53213542 on Build GRCh38
rs377159044 Chr.1: 53213602-53213602 on Build GRCh38
rs369202713 Chr.1: 53213554-53213554 on Build GRCh38
rs375766702 Chr.1: 53213519-53213519 on Build GRCh38
rs138125299 Chr.1: 53213561-53213561 on Build GRCh38
rs752325732 Chr.1: 53213394-53213395 on Build GRCh38
rs775113044 Chr.1: 53213413-53213413 on Build GRCh38
rs780286639 Chr.1: 53213440-53213440 on Build GRCh38
rs78266699 Chr.1: 53213559-53213559 on Build GRCh38
rs767004984 Chr.1: 53213392-53213393 on Build GRCh38
rs766154734 Chr.1: 53213572-53213572 on Build GRCh38
rs773979732 Chr.1: 53213494-53213494 on Build GRCh38
rs908749525 Chr.1: 53213434-53213435 on Build GRCh38
rs762495053 Chr.1: 53213486-53213486 on Build GRCh38
rs773695572 Chr.1: 53213398-53213398 on Build GRCh38
rs769535056 Chr.1: 53213475-53213475 on Build GRCh38
rs760392144 Chr.1: 53213564-53213564 on Build GRCh38
rs1040318543 Chr.1: 53213476-53213476 on Build GRCh38
rs759017526 Chr.1: 53213382-53213389 on Build GRCh38
rs757711106 Chr.1: 53213549-53213549 on Build GRCh38
rs748484701 Chr.1: 53213397-53213397 on Build GRCh38
rs756259411 Chr.1: 53213439-53213439 on Build GRCh38
rs141903761 Chr.1: 53213515-53213515 on Build GRCh38
rs112676827 Chr.1: 53213506-53213506 on Build GRCh38
rs74315293 Chr.1: 53213509-53213509 on Build GRCh38
rs768288294 Chr.1: 53213493-53213493 on Build GRCh38
rs749409744 Chr.1: 53213388-53213388 on Build GRCh38
rs903915351 Chr.1: 53213532-53213532 on Build GRCh38
rs765487506 Chr.1: 53213600-53213600 on Build GRCh38
rs747287916 Chr.1: 53213543-53213543 on Build GRCh38
rs747723963 Chr.1: 53213558-53213558 on Build GRCh38
rs28936673 Chr.1: 53213501-53213501 on Build GRCh38
rs983358783 Chr.1: 53213429-53213429 on Build GRCh38
rs540322467 Chr.1: 53213469-53213469 on Build GRCh38
rs138893547 Chr.1: 53213581-53213581 on Build GRCh38
rs1038483817 Chr.1: 53213503-53213503 on Build GRCh38
rs566533330 Chr.1: 53213483-53213483 on Build GRCh38
rs751557097 Chr.1: 53213431-53213431 on Build GRCh38
rs759970188 Chr.1: 53213490-53213490 on Build GRCh38
rs755060321 Chr.1: 53213457-53213457 on Build GRCh38
rs768470420 Chr.1: 53213556-53213556 on Build GRCh38
rs750144923 Chr.1: 53213505-53213505 on Build GRCh38
rs77565483 Chr.1: 53213385-53213385 on Build GRCh38
rs747444819 Chr.1: 53213470-53213470 on Build GRCh38
rs762584123 Chr.1: 53213418-53213418 on Build GRCh38
rs761604642 Chr.1: 53213433-53213433 on Build GRCh38
rs748596204 Chr.1: 53213460-53213460 on Build GRCh38
rs759735713 Chr.1: 53213588-53213588 on Build GRCh38
rs753861985 Chr.1: 53213441-53213441 on Build GRCh38
rs910424186 Chr.1: 53213455-53213455 on Build GRCh38
rs760255368 Chr.1: 53213400-53213400 on Build GRCh38
rs756414686 Chr.1: 53213384-53213384 on Build GRCh38
rs141146189 Chr.1: 53213394-53213394 on Build GRCh38
rs767201522 Chr.1: 53213434-53213434 on Build GRCh38
rs763889116 Chr.1: 53213502-53213502 on Build GRCh38

View Full Product Details