Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128508703-128508942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLE1
Entrez Gene ID
2733
Gene Name
GLE1, RNA export mediator
Gene Aliases
GLE1L, LCCS, LCCS1, hGLE1
UniGene
Hs.522418
Gene Chromosome Location
Chr.9: 128504692-128542301 on Build GRCh38
Transcripts
NM_001003722.1 NM_001499.2 XM_006717059.2 XM_006717060.2 XM_011518549.1 XM_011518550.1 XM_011518551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
288739 c.128C>G p.P43R Chr.9: 128508904-128508904 on Build GRCh38
5486709 c.165C>T p.H55H Chr.9: 128508941-128508941 on Build GRCh38
3847750 c.137C>T p.S46F Chr.9: 128508913-128508913 on Build GRCh38
1460269 c.164A>G p.H55R Chr.9: 128508940-128508940 on Build GRCh38
223371 c.149G>A p.G50E Chr.9: 128508925-128508925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774230809 Chr.9: 128508836-128508836 on Build GRCh38
rs1054727057 Chr.9: 128508807-128508807 on Build GRCh38
rs371811161 Chr.9: 128508775-128508775 on Build GRCh38
rs973529054 Chr.9: 128508810-128508810 on Build GRCh38
rs754271392 Chr.9: 128508853-128508853 on Build GRCh38
rs753226987 Chr.9: 128508893-128508893 on Build GRCh38
rs760803447 Chr.9: 128508848-128508848 on Build GRCh38
rs375383415 Chr.9: 128508840-128508840 on Build GRCh38
rs918716099 Chr.9: 128508758-128508758 on Build GRCh38
rs756846701 Chr.9: 128508896-128508896 on Build GRCh38
rs1015391374 Chr.9: 128508792-128508792 on Build GRCh38
rs867535211 Chr.9: 128508822-128508822 on Build GRCh38
rs972795807 Chr.9: 128508735-128508735 on Build GRCh38
rs950188082 Chr.9: 128508777-128508777 on Build GRCh38
rs766557201 Chr.9: 128508852-128508852 on Build GRCh38
rs969620233 Chr.9: 128508842-128508842 on Build GRCh38
rs927485122 Chr.9: 128508793-128508793 on Build GRCh38
rs542899467 Chr.9: 128508747-128508747 on Build GRCh38
rs962137630 Chr.9: 128508802-128508802 on Build GRCh38
rs773542362 Chr.9: 128508843-128508843 on Build GRCh38
rs749876906 Chr.9: 128508903-128508903 on Build GRCh38
rs769230078 Chr.9: 128508890-128508891 on Build GRCh38
rs759935136 Chr.9: 128508891-128508891 on Build GRCh38
rs780559879 Chr.9: 128508898-128508898 on Build GRCh38
rs755731392 Chr.9: 128508907-128508907 on Build GRCh38
rs368997996 Chr.9: 128508841-128508841 on Build GRCh38
rs893449421 Chr.9: 128508818-128508818 on Build GRCh38
rs138871311 Chr.9: 128508892-128508892 on Build GRCh38

View Full Product Details