Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 221739054-221739321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP10
Entrez Gene ID
11221
Gene Name
dual specificity phosphatase 10
Gene Aliases
MKP-5, MKP5
UniGene
Hs.497822
Gene Chromosome Location
Chr.1: 221701420-221742176 on Build GRCh38
Transcripts
NM_007207.5 NR_111940.1 XM_017000147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
239662 c.608G>A p.R203Q Chr.1: 221739137-221739137 on Build GRCh38
6650291 c.504G>A p.P168P Chr.1: 221739241-221739241 on Build GRCh38
6264305 c.603C>A p.S201R Chr.1: 221739142-221739142 on Build GRCh38
1501504 c.471G>C p.K157N Chr.1: 221739274-221739274 on Build GRCh38
6992079 c.537C>G p.P179P Chr.1: 221739208-221739208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770644727 Chr.1: 221739165-221739165 on Build GRCh38
rs142143528 Chr.1: 221739276-221739276 on Build GRCh38
rs147985011 Chr.1: 221739169-221739169 on Build GRCh38
rs375755922 Chr.1: 221739148-221739148 on Build GRCh38
rs760626501 Chr.1: 221739225-221739225 on Build GRCh38
rs777347748 Chr.1: 221739159-221739159 on Build GRCh38
rs763185891 Chr.1: 221739089-221739089 on Build GRCh38
rs750414647 Chr.1: 221739135-221739135 on Build GRCh38
rs756698798 Chr.1: 221739287-221739287 on Build GRCh38
rs903341067 Chr.1: 221739124-221739124 on Build GRCh38
rs766677636 Chr.1: 221739241-221739241 on Build GRCh38
rs775057659 Chr.1: 221739204-221739204 on Build GRCh38
rs753693790 Chr.1: 221739141-221739141 on Build GRCh38
rs746551024 Chr.1: 221739286-221739286 on Build GRCh38
rs780936253 Chr.1: 221739290-221739290 on Build GRCh38
rs746433661 Chr.1: 221739160-221739160 on Build GRCh38
rs765000706 Chr.1: 221739248-221739248 on Build GRCh38
rs767845837 Chr.1: 221739219-221739219 on Build GRCh38
rs767693304 Chr.1: 221739131-221739131 on Build GRCh38
rs74402621 Chr.1: 221739189-221739189 on Build GRCh38
rs147602708 Chr.1: 221739098-221739098 on Build GRCh38
rs370364280 Chr.1: 221739214-221739214 on Build GRCh38
rs372477751 Chr.1: 221739138-221739138 on Build GRCh38
rs374050420 Chr.1: 221739283-221739283 on Build GRCh38
rs748076536 Chr.1: 221739154-221739154 on Build GRCh38
rs138642178 Chr.1: 221739130-221739130 on Build GRCh38
rs61758448 Chr.1: 221739242-221739242 on Build GRCh38
rs775923909 Chr.1: 221739087-221739087 on Build GRCh38
rs143436731 Chr.1: 221739151-221739151 on Build GRCh38
rs769109743 Chr.1: 221739194-221739194 on Build GRCh38
rs35391196 Chr.1: 221739296-221739297 on Build GRCh38
rs766472059 Chr.1: 221739140-221739140 on Build GRCh38
rs752429127 Chr.1: 221739253-221739253 on Build GRCh38
rs772696355 Chr.1: 221739191-221739193 on Build GRCh38
rs1051946246 Chr.1: 221739136-221739136 on Build GRCh38
rs145011039 Chr.1: 221739220-221739220 on Build GRCh38
rs749716933 Chr.1: 221739190-221739190 on Build GRCh38
rs566662778 Chr.1: 221739266-221739266 on Build GRCh38
rs866230538 Chr.1: 221739203-221739203 on Build GRCh38

View Full Product Details