Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124681571-124681834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM53B
Entrez Gene ID
9679
Gene Name
family with sequence similarity 53 member B
Gene Aliases
KIAA0140, bA12J10.2, smp
UniGene
Hs.129195
Gene Chromosome Location
Chr.10: 124619294-124744361 on Build GRCh38
Transcripts
NM_014661.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1675638 c.775C>T p.R259C Chr.10: 124681738-124681738 on Build GRCh38
3768982 c.906G>A p.Q302Q Chr.10: 124681607-124681607 on Build GRCh38
1675637 c.901G>A p.A301T Chr.10: 124681612-124681612 on Build GRCh38
4413426 c.856G>A p.E286K Chr.10: 124681657-124681657 on Build GRCh38
118592 c.747A>C p.T249T Chr.10: 124681766-124681766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141674794 Chr.10: 124681703-124681703 on Build GRCh38
rs138329808 Chr.10: 124681642-124681642 on Build GRCh38
rs572365285 Chr.10: 124681743-124681743 on Build GRCh38
rs145118068 Chr.10: 124681663-124681663 on Build GRCh38
rs199729223 Chr.10: 124681690-124681690 on Build GRCh38
rs759772596 Chr.10: 124681721-124681721 on Build GRCh38
rs370313656 Chr.10: 124681691-124681691 on Build GRCh38
rs370860660 Chr.10: 124681657-124681657 on Build GRCh38
rs771291991 Chr.10: 124681791-124681791 on Build GRCh38
rs776484455 Chr.10: 124681760-124681760 on Build GRCh38
rs779974324 Chr.10: 124681757-124681757 on Build GRCh38
rs1043962767 Chr.10: 124681720-124681720 on Build GRCh38
rs539748980 Chr.10: 124681765-124681765 on Build GRCh38
rs773662292 Chr.10: 124681781-124681781 on Build GRCh38
rs756438927 Chr.10: 124681591-124681591 on Build GRCh38
rs766526096 Chr.10: 124681615-124681615 on Build GRCh38
rs747629332 Chr.10: 124681768-124681768 on Build GRCh38
rs138940439 Chr.10: 124681679-124681679 on Build GRCh38
rs1037843246 Chr.10: 124681678-124681678 on Build GRCh38
rs375676113 Chr.10: 124681804-124681804 on Build GRCh38
rs757170491 Chr.10: 124681649-124681649 on Build GRCh38
rs752778458 Chr.10: 124681730-124681730 on Build GRCh38
rs62642491 Chr.10: 124681717-124681717 on Build GRCh38
rs765399560 Chr.10: 124681729-124681729 on Build GRCh38
rs376270010 Chr.10: 124681688-124681688 on Build GRCh38
rs766731243 Chr.10: 124681713-124681713 on Build GRCh38
rs768455934 Chr.10: 124681766-124681766 on Build GRCh38
rs779639182 Chr.10: 124681661-124681661 on Build GRCh38
rs777006848 Chr.10: 124681793-124681793 on Build GRCh38
rs771601140 Chr.10: 124681774-124681774 on Build GRCh38
rs958232695 Chr.10: 124681772-124681772 on Build GRCh38
rs758446507 Chr.10: 124681737-124681737 on Build GRCh38
rs754049121 Chr.10: 124681624-124681624 on Build GRCh38
rs1032135428 Chr.10: 124681746-124681746 on Build GRCh38
rs372876962 Chr.10: 124681697-124681697 on Build GRCh38
rs761006227 Chr.10: 124681712-124681712 on Build GRCh38
rs779220632 Chr.10: 124681738-124681738 on Build GRCh38
rs761220142 Chr.10: 124681785-124681785 on Build GRCh38
rs149630052 Chr.10: 124681658-124681658 on Build GRCh38
rs765458457 Chr.10: 124681816-124681816 on Build GRCh38
rs908062845 Chr.10: 124681600-124681600 on Build GRCh38
rs932214278 Chr.10: 124681681-124681681 on Build GRCh38
rs778499544 Chr.10: 124681600-124681601 on Build GRCh38
rs746823774 Chr.10: 124681777-124681777 on Build GRCh38
rs755087032 Chr.10: 124681630-124681630 on Build GRCh38
rs781047985 Chr.10: 124681742-124681742 on Build GRCh38
rs769615824 Chr.10: 124681659-124681659 on Build GRCh38

View Full Product Details