Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9876987-9877244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIDEC
Entrez Gene ID
63924
Gene Name
cell death inducing DFFA like effector c
Gene Aliases
CIDE-3, CIDE3, FPLD5, FSP27
UniGene
Hs.567562
Gene Chromosome Location
Chr.3: 9866710-9880254 on Build GRCh38
Transcripts
NM_001199551.1 NM_001199552.1 NM_001199623.1 NM_001321142.1 NM_001321143.1 NM_001321144.1 NM_022094.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6371130 c.163A>T p.R55W Chr.3: 9877110-9877110 on Build GRCh38
447136 c.96G>T p.L32L Chr.3: 9877177-9877177 on Build GRCh38
6050962 c.159C>T p.S53S Chr.3: 9877114-9877114 on Build GRCh38
6322662 c.154C>A p.R52R Chr.3: 9877119-9877119 on Build GRCh38
6213196 c.173T>C p.I58T Chr.3: 9877100-9877100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970711982 Chr.3: 9877175-9877175 on Build GRCh38
rs776144628 Chr.3: 9877049-9877049 on Build GRCh38
rs772831652 Chr.3: 9877154-9877154 on Build GRCh38
rs199505032 Chr.3: 9877166-9877166 on Build GRCh38
rs768086965 Chr.3: 9877031-9877031 on Build GRCh38
rs375819904 Chr.3: 9877167-9877167 on Build GRCh38
rs758723139 Chr.3: 9877050-9877050 on Build GRCh38
rs61742367 Chr.3: 9877127-9877127 on Build GRCh38
rs1049018868 Chr.3: 9877224-9877224 on Build GRCh38
rs762707442 Chr.3: 9877082-9877082 on Build GRCh38
rs760803919 Chr.3: 9877176-9877176 on Build GRCh38
rs778134784 Chr.3: 9877114-9877114 on Build GRCh38
rs1003987337 Chr.3: 9877084-9877084 on Build GRCh38
rs150971509 Chr.3: 9877134-9877134 on Build GRCh38
rs752783696 Chr.3: 9877173-9877173 on Build GRCh38
rs757491895 Chr.3: 9877217-9877217 on Build GRCh38
rs148744468 Chr.3: 9877113-9877113 on Build GRCh38
rs1023533575 Chr.3: 9877164-9877164 on Build GRCh38
rs533483753 Chr.3: 9877145-9877145 on Build GRCh38
rs956988245 Chr.3: 9877137-9877137 on Build GRCh38
rs79419480 Chr.3: 9877092-9877092 on Build GRCh38
rs757670421 Chr.3: 9877119-9877119 on Build GRCh38
rs200343387 Chr.3: 9877098-9877098 on Build GRCh38
rs917890127 Chr.3: 9877157-9877157 on Build GRCh38
rs761388511 Chr.3: 9877051-9877051 on Build GRCh38
rs886844137 Chr.3: 9877067-9877067 on Build GRCh38
rs779367682 Chr.3: 9877123-9877123 on Build GRCh38
rs370392826 Chr.3: 9877136-9877136 on Build GRCh38
rs766353274 Chr.3: 9877088-9877088 on Build GRCh38
rs973565068 Chr.3: 9877029-9877029 on Build GRCh38
rs574033260 Chr.3: 9877124-9877124 on Build GRCh38
rs879198895 Chr.3: 9877102-9877102 on Build GRCh38
rs767475090 Chr.3: 9877168-9877168 on Build GRCh38
rs751474684 Chr.3: 9877090-9877090 on Build GRCh38
rs201565574 Chr.3: 9877155-9877155 on Build GRCh38
rs770743677 Chr.3: 9877160-9877160 on Build GRCh38
rs767498216 Chr.3: 9877096-9877096 on Build GRCh38
rs754291958 Chr.3: 9877118-9877118 on Build GRCh38
rs931608390 Chr.3: 9877201-9877201 on Build GRCh38
rs962039863 Chr.3: 9877215-9877215 on Build GRCh38
rs456168 Chr.3: 9877177-9877177 on Build GRCh38
rs562968333 Chr.3: 9877156-9877156 on Build GRCh38
rs988262334 Chr.3: 9877146-9877146 on Build GRCh38
rs368701651 Chr.3: 9877052-9877052 on Build GRCh38
rs773059760 Chr.3: 9877068-9877068 on Build GRCh38
rs773749639 Chr.3: 9877152-9877152 on Build GRCh38
rs931477770 Chr.3: 9877020-9877020 on Build GRCh38
rs754130497 Chr.3: 9877216-9877216 on Build GRCh38

View Full Product Details