Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62482958-62483217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO11
Entrez Gene ID
51194
Gene Name
importin 11
Gene Aliases
RanBP11
UniGene
Hs.482269
Gene Chromosome Location
Chr.5: 62412746-62628589 on Build GRCh38
Transcripts
NM_001134779.1 NM_016338.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3941351 c.957C>G p.D319E Chr.5: 62483109-62483109 on Build GRCh38
3920242 c.860C>T p.S287L Chr.5: 62483132-62483132 on Build GRCh38
3920241 c.980C>T p.S327L Chr.5: 62483132-62483132 on Build GRCh38
7263443 c.991C>A p.L331I Chr.5: 62483143-62483143 on Build GRCh38
4756292 c.1060G>A p.E354K Chr.5: 62483212-62483212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944098428 Chr.5: 62482990-62482990 on Build GRCh38
rs369750916 Chr.5: 62483082-62483082 on Build GRCh38
rs774145566 Chr.5: 62483081-62483081 on Build GRCh38
rs375679717 Chr.5: 62483145-62483145 on Build GRCh38
rs747556561 Chr.5: 62483059-62483059 on Build GRCh38
rs759164450 Chr.5: 62483090-62483093 on Build GRCh38
rs969813971 Chr.5: 62483109-62483109 on Build GRCh38
rs751548188 Chr.5: 62483154-62483154 on Build GRCh38
rs756530541 Chr.5: 62483182-62483182 on Build GRCh38
rs772526004 Chr.5: 62483096-62483096 on Build GRCh38
rs895588286 Chr.5: 62483009-62483009 on Build GRCh38
rs372627658 Chr.5: 62483093-62483093 on Build GRCh38
rs752931991 Chr.5: 62483140-62483140 on Build GRCh38
rs781639874 Chr.5: 62483069-62483069 on Build GRCh38
rs769489452 Chr.5: 62483090-62483090 on Build GRCh38
rs954847590 Chr.5: 62483177-62483177 on Build GRCh38
rs998069939 Chr.5: 62483156-62483156 on Build GRCh38
rs983388401 Chr.5: 62483192-62483192 on Build GRCh38
rs781523245 Chr.5: 62483169-62483169 on Build GRCh38
rs546633931 Chr.5: 62483048-62483048 on Build GRCh38
rs771718475 Chr.5: 62483083-62483083 on Build GRCh38
rs1000116544 Chr.5: 62483062-62483062 on Build GRCh38
rs746305183 Chr.5: 62483174-62483174 on Build GRCh38
rs757835275 Chr.5: 62483161-62483161 on Build GRCh38
rs539500243 Chr.5: 62483019-62483019 on Build GRCh38
rs945688372 Chr.5: 62483010-62483010 on Build GRCh38
rs768466231 Chr.5: 62483080-62483080 on Build GRCh38
rs901653048 Chr.5: 62483039-62483039 on Build GRCh38
rs759294534 Chr.5: 62483135-62483135 on Build GRCh38
rs771304234 Chr.5: 62483068-62483068 on Build GRCh38
rs368152207 Chr.5: 62483136-62483136 on Build GRCh38
rs746200518 Chr.5: 62483071-62483071 on Build GRCh38
rs566434659 Chr.5: 62483049-62483049 on Build GRCh38
rs776537031 Chr.5: 62483122-62483122 on Build GRCh38
rs766178169 Chr.5: 62483120-62483120 on Build GRCh38
rs1041414746 Chr.5: 62483025-62483025 on Build GRCh38
rs760697610 Chr.5: 62483099-62483099 on Build GRCh38
rs772596657 Chr.5: 62483078-62483081 on Build GRCh38
rs1022583951 Chr.5: 62483095-62483095 on Build GRCh38
rs372051641 Chr.5: 62483142-62483142 on Build GRCh38

View Full Product Details