Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48033132-48033389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPZ2
Entrez Gene ID
51226
Gene Name
coatomer protein complex subunit zeta 2
Gene Aliases
zeta2-COP
UniGene
Hs.408434
Gene Chromosome Location
Chr.17: 48026167-48048091 on Build GRCh38
Transcripts
NM_016429.3 XM_011524874.2

SNP Detail

SNP ID Chromosome Location
rs780849615 Chr.17: 48033286-48033286 on Build GRCh38
rs564567275 Chr.17: 48033216-48033216 on Build GRCh38
rs1041394766 Chr.17: 48033228-48033228 on Build GRCh38
rs546983859 Chr.17: 48033254-48033254 on Build GRCh38
rs866362844 Chr.17: 48033270-48033270 on Build GRCh38
rs756855561 Chr.17: 48033193-48033193 on Build GRCh38
rs752487176 Chr.17: 48033249-48033249 on Build GRCh38
rs777462841 Chr.17: 48033188-48033188 on Build GRCh38
rs1004515887 Chr.17: 48033361-48033361 on Build GRCh38
rs763753449 Chr.17: 48033275-48033275 on Build GRCh38
rs769137167 Chr.17: 48033201-48033201 on Build GRCh38
rs779450888 Chr.17: 48033310-48033310 on Build GRCh38
rs368441108 Chr.17: 48033340-48033340 on Build GRCh38
rs951345414 Chr.17: 48033354-48033354 on Build GRCh38
rs372127390 Chr.17: 48033281-48033281 on Build GRCh38
rs777421784 Chr.17: 48033255-48033255 on Build GRCh38
rs968286923 Chr.17: 48033169-48033169 on Build GRCh38
rs577050882 Chr.17: 48033181-48033181 on Build GRCh38
rs773451971 Chr.17: 48033227-48033227 on Build GRCh38
rs998015526 Chr.17: 48033259-48033259 on Build GRCh38
rs756908411 Chr.17: 48033284-48033284 on Build GRCh38
rs375107752 Chr.17: 48033328-48033328 on Build GRCh38
rs765212190 Chr.17: 48033246-48033246 on Build GRCh38
rs371433033 Chr.17: 48033237-48033237 on Build GRCh38
rs201805900 Chr.17: 48033220-48033220 on Build GRCh38
rs755495957 Chr.17: 48033308-48033308 on Build GRCh38
rs374731995 Chr.17: 48033238-48033238 on Build GRCh38
rs776752998 Chr.17: 48033330-48033330 on Build GRCh38
rs748531919 Chr.17: 48033219-48033219 on Build GRCh38
rs764947412 Chr.17: 48033171-48033171 on Build GRCh38
rs948624910 Chr.17: 48033326-48033326 on Build GRCh38
rs748500086 Chr.17: 48033318-48033318 on Build GRCh38
rs758048951 Chr.17: 48033184-48033184 on Build GRCh38
rs749947755 Chr.17: 48033296-48033296 on Build GRCh38
rs551140711 Chr.17: 48033198-48033198 on Build GRCh38
rs927495851 Chr.17: 48033159-48033159 on Build GRCh38
rs780798202 Chr.17: 48033197-48033197 on Build GRCh38
rs772510371 Chr.17: 48033320-48033320 on Build GRCh38
rs375665368 Chr.17: 48033234-48033234 on Build GRCh38
rs112698182 Chr.17: 48033210-48033210 on Build GRCh38
rs1003060022 Chr.17: 48033329-48033329 on Build GRCh38
rs759539918 Chr.17: 48033239-48033239 on Build GRCh38
rs937503582 Chr.17: 48033170-48033170 on Build GRCh38
rs530549189 Chr.17: 48033190-48033190 on Build GRCh38
rs778055906 Chr.17: 48033327-48033327 on Build GRCh38

View Full Product Details