Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70733580-70733853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K9
Entrez Gene ID
4293
Gene Name
mitogen-activated protein kinase kinase kinase 9
Gene Aliases
MEKK9, MLK1, PRKE1
UniGene
Hs.445496
Gene Chromosome Location
Chr.14: 70722526-70809268 on Build GRCh38
Transcripts
NM_001284230.1 NM_001284231.1 NM_001284232.1 NM_033141.3 XM_005267683.4 XM_011536788.2 XM_011536792.1 XM_011536794.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1370856 c.2059G>A p.D687N Chr.14: 70733766-70733766 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375103723 Chr.14: 70733709-70733709 on Build GRCh38
rs182530403 Chr.14: 70733724-70733724 on Build GRCh38
rs964304820 Chr.14: 70733782-70733782 on Build GRCh38
rs73282810 Chr.14: 70733727-70733727 on Build GRCh38
rs773762945 Chr.14: 70733766-70733766 on Build GRCh38
rs1018716893 Chr.14: 70733768-70733768 on Build GRCh38
rs541965205 Chr.14: 70733767-70733767 on Build GRCh38
rs373932646 Chr.14: 70733785-70733785 on Build GRCh38
rs543299403 Chr.14: 70733700-70733700 on Build GRCh38
rs780322179 Chr.14: 70733721-70733721 on Build GRCh38
rs1038142680 Chr.14: 70733671-70733671 on Build GRCh38
rs747553209 Chr.14: 70733776-70733776 on Build GRCh38
rs575973695 Chr.14: 70733653-70733653 on Build GRCh38
rs931477694 Chr.14: 70733792-70733792 on Build GRCh38
rs1001429933 Chr.14: 70733608-70733608 on Build GRCh38
rs907549967 Chr.14: 70733616-70733616 on Build GRCh38
rs772378118 Chr.14: 70733762-70733762 on Build GRCh38
rs778543493 Chr.14: 70733720-70733720 on Build GRCh38
rs779035511 Chr.14: 70733737-70733737 on Build GRCh38
rs746369819 Chr.14: 70733816-70733817 on Build GRCh38
rs748486765 Chr.14: 70733740-70733740 on Build GRCh38

View Full Product Details