Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176093291-176093530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD13
Entrez Gene ID
3239
Gene Name
homeobox D13
Gene Aliases
BDE, BDSD, HOX4I, SPD, SPD1
UniGene
Hs.152414
Gene Chromosome Location
Chr.2: 176087487-176095938 on Build GRCh38
Transcripts
NM_000523.3 XM_011511068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3668849 c.490G>A p.V164M Chr.2: 176093380-176093380 on Build GRCh38
7098991 c.609G>T p.V203V Chr.2: 176093499-176093499 on Build GRCh38
5674326 c.480A>T p.G160G Chr.2: 176093370-176093370 on Build GRCh38
3570173 c.469G>A p.A157T Chr.2: 176093359-176093359 on Build GRCh38
6026436 c.586C>T p.P196S Chr.2: 176093501-176093501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761102776 Chr.2: 176093458-176093458 on Build GRCh38
rs140031618 Chr.2: 176093316-176093316 on Build GRCh38
rs536676052 Chr.2: 176093452-176093452 on Build GRCh38
rs779934918 Chr.2: 176093495-176093495 on Build GRCh38
rs769954517 Chr.2: 176093319-176093319 on Build GRCh38
rs756710908 Chr.2: 176093392-176093392 on Build GRCh38
rs566987103 Chr.2: 176093505-176093505 on Build GRCh38
rs745798601 Chr.2: 176093410-176093410 on Build GRCh38
rs774419506 Chr.2: 176093449-176093449 on Build GRCh38
rs755848849 Chr.2: 176093380-176093380 on Build GRCh38
rs746854273 Chr.2: 176093503-176093503 on Build GRCh38
rs922763007 Chr.2: 176093446-176093446 on Build GRCh38
rs187698441 Chr.2: 176093359-176093359 on Build GRCh38
rs554977181 Chr.2: 176093489-176093489 on Build GRCh38
rs753431560 Chr.2: 176093466-176093466 on Build GRCh38
rs201904809 Chr.2: 176093423-176093423 on Build GRCh38
rs756869286 Chr.2: 176093469-176093469 on Build GRCh38
rs771939727 Chr.2: 176093411-176093411 on Build GRCh38
rs1018548542 Chr.2: 176093339-176093339 on Build GRCh38
rs778978474 Chr.2: 176093397-176093397 on Build GRCh38
rs773372452 Chr.2: 176093323-176093323 on Build GRCh38
rs762909441 Chr.2: 176093325-176093325 on Build GRCh38
rs771851753 Chr.2: 176093312-176093312 on Build GRCh38
rs148979353 Chr.2: 176093431-176093431 on Build GRCh38
rs147720746 Chr.2: 176093507-176093507 on Build GRCh38
rs751994842 Chr.2: 176093351-176093351 on Build GRCh38
rs753157172 Chr.2: 176093455-176093455 on Build GRCh38
rs779846895 Chr.2: 176093417-176093417 on Build GRCh38
rs760041709 Chr.2: 176093354-176093354 on Build GRCh38
rs13392701 Chr.2: 176093433-176093433 on Build GRCh38
rs546802875 Chr.2: 176093322-176093322 on Build GRCh38
rs766957833 Chr.2: 176093334-176093334 on Build GRCh38
rs374086950 Chr.2: 176093479-176093479 on Build GRCh38
rs758363080 Chr.2: 176093492-176093492 on Build GRCh38
rs777264698 Chr.2: 176093382-176093382 on Build GRCh38
rs200067774 Chr.2: 176093390-176093390 on Build GRCh38
rs768084107 Chr.2: 176093453-176093453 on Build GRCh38
rs775201150 Chr.2: 176093313-176093313 on Build GRCh38
rs371806521 Chr.2: 176093364-176093364 on Build GRCh38
rs773282226 Chr.2: 176093434-176093434 on Build GRCh38
rs763867556 Chr.2: 176093460-176093460 on Build GRCh38
rs753049145 Chr.2: 176093360-176093360 on Build GRCh38
rs1047620716 Chr.2: 176093500-176093500 on Build GRCh38
rs762569134 Chr.2: 176093379-176093379 on Build GRCh38
rs763149315 Chr.2: 176093447-176093447 on Build GRCh38

View Full Product Details