Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52237024-52237253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17F
Entrez Gene ID
112744
Gene Name
interleukin 17F
Gene Aliases
CANDF6, IL-17F, ML-1, ML1
UniGene
Hs.272295
Gene Chromosome Location
Chr.6: 52236681-52245689 on Build GRCh38
Transcripts
NM_052872.3 XM_011514276.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
77885 c.278A>T p.Y93F Chr.6: 52237145-52237145 on Build GRCh38
1644671 c.277T>C p.Y93H Chr.6: 52237146-52237146 on Build GRCh38
3354980 c.387C>T p.V129V Chr.6: 52237036-52237036 on Build GRCh38
5405564 c.393G>A p.R131R Chr.6: 52237030-52237030 on Build GRCh38
300068 c.383T>C p.L128P Chr.6: 52237040-52237040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972404643 Chr.6: 52237173-52237173 on Build GRCh38
rs369091221 Chr.6: 52237180-52237180 on Build GRCh38
rs776587391 Chr.6: 52237101-52237101 on Build GRCh38
rs756602791 Chr.6: 52237147-52237147 on Build GRCh38
rs901898355 Chr.6: 52237104-52237104 on Build GRCh38
rs137981743 Chr.6: 52237078-52237078 on Build GRCh38
rs751869421 Chr.6: 52237211-52237211 on Build GRCh38
rs758436603 Chr.6: 52237110-52237110 on Build GRCh38
rs113299618 Chr.6: 52237171-52237171 on Build GRCh38
rs751817035 Chr.6: 52237120-52237120 on Build GRCh38
rs781668456 Chr.6: 52237138-52237138 on Build GRCh38
rs756572344 Chr.6: 52237209-52237209 on Build GRCh38
rs1048156234 Chr.6: 52237214-52237214 on Build GRCh38
rs766543490 Chr.6: 52237207-52237207 on Build GRCh38
rs750479457 Chr.6: 52237109-52237109 on Build GRCh38
rs1029127897 Chr.6: 52237133-52237133 on Build GRCh38
rs550486674 Chr.6: 52237062-52237062 on Build GRCh38
rs2397084 Chr.6: 52237046-52237046 on Build GRCh38
rs376780230 Chr.6: 52237148-52237148 on Build GRCh38
rs752788417 Chr.6: 52237219-52237219 on Build GRCh38
rs775184787 Chr.6: 52237150-52237150 on Build GRCh38
rs556851049 Chr.6: 52237232-52237232 on Build GRCh38
rs760533244 Chr.6: 52237096-52237096 on Build GRCh38
rs748486078 Chr.6: 52237139-52237139 on Build GRCh38
rs775060397 Chr.6: 52237080-52237080 on Build GRCh38
rs773196295 Chr.6: 52237197-52237197 on Build GRCh38
rs937217588 Chr.6: 52237042-52237042 on Build GRCh38
rs767452983 Chr.6: 52237073-52237073 on Build GRCh38
rs780259967 Chr.6: 52237043-52237043 on Build GRCh38
rs762999820 Chr.6: 52237203-52237203 on Build GRCh38
rs766577007 Chr.6: 52237115-52237115 on Build GRCh38
rs755441459 Chr.6: 52237130-52237130 on Build GRCh38
rs755388522 Chr.6: 52237048-52237048 on Build GRCh38
rs148940532 Chr.6: 52237105-52237105 on Build GRCh38
rs768255263 Chr.6: 52237159-52237159 on Build GRCh38
rs763865065 Chr.6: 52237099-52237099 on Build GRCh38
rs533677359 Chr.6: 52237058-52237058 on Build GRCh38
rs566180562 Chr.6: 52237231-52237231 on Build GRCh38
rs200163061 Chr.6: 52237114-52237114 on Build GRCh38
rs570911210 Chr.6: 52237055-52237055 on Build GRCh38
rs971133931 Chr.6: 52237223-52237223 on Build GRCh38
rs146083682 Chr.6: 52237107-52237107 on Build GRCh38
rs753016908 Chr.6: 52237220-52237220 on Build GRCh38
rs1042175463 Chr.6: 52237076-52237076 on Build GRCh38
rs761606944 Chr.6: 52237181-52237181 on Build GRCh38
rs765086960 Chr.6: 52237190-52237190 on Build GRCh38
rs142962486 Chr.6: 52237149-52237149 on Build GRCh38
rs536968854 Chr.6: 52237063-52237063 on Build GRCh38
rs758566976 Chr.6: 52237041-52237041 on Build GRCh38
rs746645424 Chr.6: 52237153-52237153 on Build GRCh38
rs868780060 Chr.6: 52237053-52237053 on Build GRCh38
rs759871664 Chr.6: 52237212-52237212 on Build GRCh38
rs374838092 Chr.6: 52237177-52237177 on Build GRCh38

View Full Product Details