Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3013451-3013676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN9
Entrez Gene ID
9080
Gene Name
claudin 9
Gene Aliases
-
UniGene
Hs.296949
Gene Chromosome Location
Chr.16: 3012456-3014505 on Build GRCh38
Transcripts
NM_020982.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3112542 c.116G>A p.S39N Chr.16: 3013478-3013478 on Build GRCh38
1629869 c.250T>C p.C84R Chr.16: 3013612-3013612 on Build GRCh38
6630060 c.109G>A p.G37S Chr.16: 3013471-3013471 on Build GRCh38
4532604 c.189G>T p.Q63H Chr.16: 3013551-3013551 on Build GRCh38
1293567 c.267_268GC>TG p.L90V Chr.16: 3013629-3013630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142749972 Chr.16: 3013525-3013525 on Build GRCh38
rs77321442 Chr.16: 3013583-3013583 on Build GRCh38
rs372231955 Chr.16: 3013539-3013539 on Build GRCh38
rs748653676 Chr.16: 3013599-3013599 on Build GRCh38
rs775175337 Chr.16: 3013604-3013604 on Build GRCh38
rs753769971 Chr.16: 3013568-3013568 on Build GRCh38
rs868531041 Chr.16: 3013645-3013645 on Build GRCh38
rs762719346 Chr.16: 3013488-3013488 on Build GRCh38
rs755940060 Chr.16: 3013510-3013510 on Build GRCh38
rs1022400616 Chr.16: 3013647-3013647 on Build GRCh38
rs763070067 Chr.16: 3013632-3013632 on Build GRCh38
rs372118474 Chr.16: 3013619-3013619 on Build GRCh38
rs761935222 Chr.16: 3013569-3013569 on Build GRCh38
rs750660041 Chr.16: 3013578-3013578 on Build GRCh38
rs757420145 Chr.16: 3013469-3013469 on Build GRCh38
rs867461598 Chr.16: 3013553-3013553 on Build GRCh38
rs779085754 Chr.16: 3013471-3013471 on Build GRCh38
rs770579664 Chr.16: 3013600-3013600 on Build GRCh38
rs145882247 Chr.16: 3013483-3013483 on Build GRCh38
rs747585456 Chr.16: 3013533-3013533 on Build GRCh38
rs201252752 Chr.16: 3013520-3013520 on Build GRCh38
rs1009684231 Chr.16: 3013638-3013638 on Build GRCh38
rs138697130 Chr.16: 3013506-3013506 on Build GRCh38
rs201203260 Chr.16: 3013618-3013618 on Build GRCh38
rs759442841 Chr.16: 3013499-3013499 on Build GRCh38
rs746072973 Chr.16: 3013523-3013523 on Build GRCh38
rs574035742 Chr.16: 3013538-3013538 on Build GRCh38
rs202102733 Chr.16: 3013540-3013540 on Build GRCh38
rs772316258 Chr.16: 3013473-3013473 on Build GRCh38
rs375644575 Chr.16: 3013603-3013603 on Build GRCh38
rs765328047 Chr.16: 3013624-3013624 on Build GRCh38
rs760454690 Chr.16: 3013606-3013606 on Build GRCh38
rs554077504 Chr.16: 3013535-3013535 on Build GRCh38
rs772812332 Chr.16: 3013486-3013486 on Build GRCh38
rs764058885 Chr.16: 3013567-3013567 on Build GRCh38
rs769348025 Chr.16: 3013485-3013485 on Build GRCh38
rs755451876 Chr.16: 3013646-3013646 on Build GRCh38
rs765599212 Chr.16: 3013532-3013532 on Build GRCh38
rs544084273 Chr.16: 3013588-3013588 on Build GRCh38
rs142023644 Chr.16: 3013651-3013651 on Build GRCh38
rs1044675110 Chr.16: 3013597-3013597 on Build GRCh38
rs200358793 Chr.16: 3013642-3013642 on Build GRCh38
rs151129127 Chr.16: 3013584-3013584 on Build GRCh38
rs781642627 Chr.16: 3013649-3013649 on Build GRCh38
rs778320259 Chr.16: 3013654-3013654 on Build GRCh38
rs760494708 Chr.16: 3013564-3013564 on Build GRCh38
rs200336281 Chr.16: 3013470-3013470 on Build GRCh38
rs374545504 Chr.16: 3013633-3013633 on Build GRCh38
rs748929677 Chr.16: 3013513-3013514 on Build GRCh38
rs781782556 Chr.16: 3013593-3013593 on Build GRCh38
rs750418823 Chr.16: 3013625-3013625 on Build GRCh38
rs374973139 Chr.16: 3013563-3013563 on Build GRCh38
rs145786872 Chr.16: 3013482-3013482 on Build GRCh38
rs746271447 Chr.16: 3013472-3013472 on Build GRCh38
rs758798960 Chr.16: 3013524-3013524 on Build GRCh38
rs765239666 Chr.16: 3013572-3013572 on Build GRCh38
rs766188648 Chr.16: 3013497-3013497 on Build GRCh38
rs770737027 Chr.16: 3013537-3013538 on Build GRCh38
rs767203719 Chr.16: 3013503-3013503 on Build GRCh38
rs768426809 Chr.16: 3013613-3013613 on Build GRCh38
rs764180087 Chr.16: 3013514-3013514 on Build GRCh38
rs147520555 Chr.16: 3013582-3013582 on Build GRCh38
rs774119241 Chr.16: 3013498-3013498 on Build GRCh38
rs756651062 Chr.16: 3013652-3013652 on Build GRCh38
rs779172380 Chr.16: 3013521-3013521 on Build GRCh38
rs767350382 Chr.16: 3013542-3013542 on Build GRCh38

View Full Product Details