Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2663463-2663732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH3B
Entrez Gene ID
3420
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) beta
Gene Aliases
RP46
UniGene
Hs.436405
Gene Chromosome Location
Chr.20: 2658390-2664223 on Build GRCh38
Transcripts
NM_001258384.2 NM_006899.4 NM_174855.3 NR_136344.1 XM_005260716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6683634 c.265C>T p.Q89* Chr.20: 2663518-2663518 on Build GRCh38
6683635 c.265C>T p.Q89* Chr.20: 2663518-2663518 on Build GRCh38
5391746 c.149C>T p.P50L Chr.20: 2663727-2663727 on Build GRCh38
5391747 c.149C>T p.P50L Chr.20: 2663727-2663727 on Build GRCh38
4813761 c.175G>T p.V59L Chr.20: 2663701-2663701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769737054 Chr.20: 2663562-2663562 on Build GRCh38
rs1029862245 Chr.20: 2663634-2663634 on Build GRCh38
rs369809528 Chr.20: 2663510-2663510 on Build GRCh38
rs770148916 Chr.20: 2663680-2663680 on Build GRCh38
rs139159095 Chr.20: 2663531-2663531 on Build GRCh38
rs772312939 Chr.20: 2663579-2663579 on Build GRCh38
rs747126644 Chr.20: 2663649-2663649 on Build GRCh38
rs757925546 Chr.20: 2663530-2663530 on Build GRCh38
rs771116491 Chr.20: 2663596-2663596 on Build GRCh38
rs760965916 Chr.20: 2663592-2663592 on Build GRCh38
rs759903876 Chr.20: 2663673-2663673 on Build GRCh38
rs376130482 Chr.20: 2663632-2663632 on Build GRCh38
rs781026048 Chr.20: 2663558-2663558 on Build GRCh38
rs775620619 Chr.20: 2663574-2663574 on Build GRCh38
rs373677018 Chr.20: 2663619-2663619 on Build GRCh38
rs370462986 Chr.20: 2663650-2663650 on Build GRCh38
rs748822744 Chr.20: 2663527-2663528 on Build GRCh38
rs752179858 Chr.20: 2663529-2663529 on Build GRCh38
rs765800635 Chr.20: 2663522-2663522 on Build GRCh38
rs753232330 Chr.20: 2663609-2663609 on Build GRCh38
rs940426994 Chr.20: 2663519-2663519 on Build GRCh38
rs777986948 Chr.20: 2663648-2663648 on Build GRCh38
rs749245355 Chr.20: 2663647-2663647 on Build GRCh38
rs1001138607 Chr.20: 2663627-2663627 on Build GRCh38
rs748194013 Chr.20: 2663575-2663575 on Build GRCh38
rs756928303 Chr.20: 2663635-2663635 on Build GRCh38
rs764648168 Chr.20: 2663704-2663704 on Build GRCh38
rs375803338 Chr.20: 2663711-2663711 on Build GRCh38
rs754242405 Chr.20: 2663504-2663504 on Build GRCh38
rs745712607 Chr.20: 2663561-2663561 on Build GRCh38
rs372837005 Chr.20: 2663689-2663689 on Build GRCh38
rs777051219 Chr.20: 2663598-2663598 on Build GRCh38
rs751084358 Chr.20: 2663625-2663625 on Build GRCh38
rs756941685 Chr.20: 2663543-2663543 on Build GRCh38
rs199958758 Chr.20: 2663595-2663595 on Build GRCh38
rs780745599 Chr.20: 2663638-2663638 on Build GRCh38
rs780103581 Chr.20: 2663645-2663645 on Build GRCh38
rs1023596307 Chr.20: 2663539-2663539 on Build GRCh38
rs765682076 Chr.20: 2663607-2663607 on Build GRCh38
rs201186040 Chr.20: 2663618-2663618 on Build GRCh38
rs759860011 Chr.20: 2663601-2663601 on Build GRCh38
rs779486676 Chr.20: 2663498-2663500 on Build GRCh38
rs1036032788 Chr.20: 2663530-2663532 on Build GRCh38
rs1013250648 Chr.20: 2663538-2663538 on Build GRCh38
rs776739072 Chr.20: 2663657-2663657 on Build GRCh38
rs773504379 Chr.20: 2663590-2663590 on Build GRCh38
rs755998017 Chr.20: 2663639-2663639 on Build GRCh38
rs746677639 Chr.20: 2663542-2663542 on Build GRCh38
rs775917325 Chr.20: 2663686-2663686 on Build GRCh38

View Full Product Details