Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50843601-50843846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEU7
Entrez Gene ID
220107
Gene Name
deleted in lymphocytic leukemia, 7
Gene Aliases
-
UniGene
Hs.673860
Gene Chromosome Location
Chr.13: 50711026-50843939 on Build GRCh38
Transcripts
NM_001306135.1 NM_198989.3 NR_046551.1

Gene Symbol
DLEU7-AS1
Entrez Gene ID
100874074
Gene Name
DLEU7 antisense RNA 1
Gene Aliases
-
UniGene
Hs.734030
Gene Chromosome Location
Chr.13: 50807855-50849905 on Build GRCh38
Transcripts
NM_001306135.1 NM_198989.3 NR_046551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398847 c.28T>C p.S10P Chr.13: 50843619-50843619 on Build GRCh38
4621595 c.6C>A p.A2A Chr.13: 50843641-50843641 on Build GRCh38
6398846 c.28T>C p.S10P Chr.13: 50843619-50843619 on Build GRCh38
6360758 c.7A>G p.S3G Chr.13: 50843640-50843640 on Build GRCh38
6360757 c.7A>G p.S3G Chr.13: 50843640-50843640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574625481 Chr.13: 50843716-50843716 on Build GRCh38
rs564420556 Chr.13: 50843812-50843812 on Build GRCh38
rs541245998 Chr.13: 50843664-50843664 on Build GRCh38
rs773005992 Chr.13: 50843696-50843696 on Build GRCh38
rs959105220 Chr.13: 50843817-50843817 on Build GRCh38
rs758364109 Chr.13: 50843813-50843813 on Build GRCh38
rs748081839 Chr.13: 50843668-50843668 on Build GRCh38
rs553127109 Chr.13: 50843709-50843709 on Build GRCh38
rs571242579 Chr.13: 50843622-50843622 on Build GRCh38
rs1054734908 Chr.13: 50843770-50843770 on Build GRCh38
rs920294747 Chr.13: 50843667-50843667 on Build GRCh38
rs1041785301 Chr.13: 50843766-50843766 on Build GRCh38
rs1040267421 Chr.13: 50843688-50843688 on Build GRCh38
rs1051080003 Chr.13: 50843741-50843741 on Build GRCh38
rs771770695 Chr.13: 50843684-50843684 on Build GRCh38
rs530936420 Chr.13: 50843806-50843806 on Build GRCh38
rs545837511 Chr.13: 50843808-50843808 on Build GRCh38
rs147380577 Chr.13: 50843750-50843750 on Build GRCh38
rs998770473 Chr.13: 50843752-50843752 on Build GRCh38
rs1002904470 Chr.13: 50843791-50843791 on Build GRCh38
rs769429571 Chr.13: 50843659-50843659 on Build GRCh38
rs983232414 Chr.13: 50843671-50843671 on Build GRCh38
rs959020372 Chr.13: 50843658-50843658 on Build GRCh38
rs541910127 Chr.13: 50843747-50843747 on Build GRCh38
rs907335556 Chr.13: 50843771-50843771 on Build GRCh38
rs774247321 Chr.13: 50843665-50843665 on Build GRCh38
rs907588941 Chr.13: 50843676-50843676 on Build GRCh38
rs566478487 Chr.13: 50843774-50843774 on Build GRCh38
rs77540621 Chr.13: 50843663-50843663 on Build GRCh38
rs777208121 Chr.13: 50843705-50843705 on Build GRCh38
rs892962951 Chr.13: 50843772-50843772 on Build GRCh38
rs893150761 Chr.13: 50843746-50843746 on Build GRCh38
rs868180599 Chr.13: 50843654-50843654 on Build GRCh38
rs779597039 Chr.13: 50843660-50843660 on Build GRCh38
rs899886073 Chr.13: 50843693-50843693 on Build GRCh38
rs931943843 Chr.13: 50843739-50843739 on Build GRCh38

View Full Product Details