Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110647016-110647265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARS2
Entrez Gene ID
79587
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD27, cysRS
UniGene
Hs.508725
Gene Chromosome Location
Chr.13: 110641410-110713527 on Build GRCh38
Transcripts
NM_024537.3 XM_006719953.2 XM_011521116.1 XM_017020741.1 XM_017020742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4817800 c.1143G>A p.M381I Chr.13: 110647151-110647151 on Build GRCh38
7295136 c.1116T>A p.S372S Chr.13: 110647178-110647178 on Build GRCh38
5478445 c.1089T>C p.A363A Chr.13: 110647205-110647205 on Build GRCh38
1188746 c.1147G>T p.G383W Chr.13: 110647147-110647147 on Build GRCh38
6408456 c.1143G>T p.M381I Chr.13: 110647151-110647151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028758261 Chr.13: 110647039-110647039 on Build GRCh38
rs770452008 Chr.13: 110647227-110647227 on Build GRCh38
rs747585829 Chr.13: 110647216-110647216 on Build GRCh38
rs200079151 Chr.13: 110647235-110647235 on Build GRCh38
rs770630631 Chr.13: 110647161-110647161 on Build GRCh38
rs551971117 Chr.13: 110647208-110647208 on Build GRCh38
rs547702190 Chr.13: 110647165-110647165 on Build GRCh38
rs113959865 Chr.13: 110647068-110647068 on Build GRCh38
rs745633983 Chr.13: 110647233-110647233 on Build GRCh38
rs761332389 Chr.13: 110647075-110647075 on Build GRCh38
rs750743449 Chr.13: 110647183-110647183 on Build GRCh38
rs201421653 Chr.13: 110647079-110647079 on Build GRCh38
rs138372705 Chr.13: 110647126-110647126 on Build GRCh38
rs568277703 Chr.13: 110647229-110647229 on Build GRCh38
rs780567107 Chr.13: 110647135-110647135 on Build GRCh38
rs748852872 Chr.13: 110647224-110647224 on Build GRCh38
rs771260058 Chr.13: 110647242-110647244 on Build GRCh38
rs779076277 Chr.13: 110647128-110647128 on Build GRCh38
rs370357923 Chr.13: 110647093-110647093 on Build GRCh38
rs185149092 Chr.13: 110647076-110647076 on Build GRCh38
rs545957777 Chr.13: 110647115-110647115 on Build GRCh38
rs148857540 Chr.13: 110647153-110647153 on Build GRCh38
rs756299705 Chr.13: 110647116-110647116 on Build GRCh38
rs41275134 Chr.13: 110647127-110647127 on Build GRCh38
rs79081036 Chr.13: 110647162-110647162 on Build GRCh38
rs765515018 Chr.13: 110647182-110647182 on Build GRCh38
rs143638615 Chr.13: 110647220-110647220 on Build GRCh38
rs747637203 Chr.13: 110647146-110647146 on Build GRCh38
rs1030991366 Chr.13: 110647187-110647187 on Build GRCh38
rs749680674 Chr.13: 110647225-110647226 on Build GRCh38
rs368032057 Chr.13: 110647089-110647089 on Build GRCh38
rs972372424 Chr.13: 110647119-110647119 on Build GRCh38
rs200826696 Chr.13: 110647223-110647223 on Build GRCh38
rs142070578 Chr.13: 110647166-110647166 on Build GRCh38
rs893366940 Chr.13: 110647211-110647211 on Build GRCh38
rs757359798 Chr.13: 110647177-110647177 on Build GRCh38
rs894080442 Chr.13: 110647188-110647188 on Build GRCh38
rs759437928 Chr.13: 110647163-110647163 on Build GRCh38
rs527311442 Chr.13: 110647193-110647193 on Build GRCh38
rs961910393 Chr.13: 110647087-110647087 on Build GRCh38
rs753952896 Chr.13: 110647176-110647176 on Build GRCh38
rs764146777 Chr.13: 110647167-110647167 on Build GRCh38
rs543658268 Chr.13: 110647152-110647152 on Build GRCh38
rs772918047 Chr.13: 110647154-110647154 on Build GRCh38
rs570568500 Chr.13: 110647219-110647219 on Build GRCh38
rs955400680 Chr.13: 110647049-110647049 on Build GRCh38
rs772515282 Chr.13: 110647133-110647133 on Build GRCh38
rs1049713891 Chr.13: 110647237-110647237 on Build GRCh38
rs868498923 Chr.13: 110647117-110647117 on Build GRCh38
rs748825203 Chr.13: 110647159-110647159 on Build GRCh38
rs751374024 Chr.13: 110647080-110647080 on Build GRCh38
rs148620239 Chr.13: 110647078-110647078 on Build GRCh38
rs746241006 Chr.13: 110647132-110647132 on Build GRCh38
rs772568403 Chr.13: 110647064-110647064 on Build GRCh38
rs930563605 Chr.13: 110647113-110647113 on Build GRCh38

View Full Product Details