Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35078916-35079143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM177A1
Entrez Gene ID
283635
Gene Name
family with sequence similarity 177 member A1
Gene Aliases
C14orf24
UniGene
Hs.446357
Gene Chromosome Location
Chr.14: 35044907-35083383 on Build GRCh38
Transcripts
NM_001079519.1 NM_001289022.1 NM_173607.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658951 c.491G>A p.R164Q Chr.14: 35079011-35079011 on Build GRCh38
5694650 c.431A>C p.K144T Chr.14: 35079020-35079020 on Build GRCh38
5647279 c.437C>T p.S146F Chr.14: 35078957-35078957 on Build GRCh38
4836277 c.437C>G p.S146C Chr.14: 35078957-35078957 on Build GRCh38
6658952 c.422G>A p.R141Q Chr.14: 35079011-35079011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372726740 Chr.14: 35079027-35079027 on Build GRCh38
rs199772768 Chr.14: 35079040-35079040 on Build GRCh38
rs956067692 Chr.14: 35079088-35079088 on Build GRCh38
rs763116779 Chr.14: 35079009-35079009 on Build GRCh38
rs183647679 Chr.14: 35079068-35079068 on Build GRCh38
rs1027197919 Chr.14: 35079012-35079012 on Build GRCh38
rs996146905 Chr.14: 35079010-35079010 on Build GRCh38
rs763303368 Chr.14: 35079011-35079011 on Build GRCh38
rs970463256 Chr.14: 35079101-35079101 on Build GRCh38
rs556524369 Chr.14: 35079019-35079019 on Build GRCh38
rs764083493 Chr.14: 35078991-35078991 on Build GRCh38
rs374405806 Chr.14: 35079035-35079035 on Build GRCh38
rs567166491 Chr.14: 35078962-35078962 on Build GRCh38
rs145536297 Chr.14: 35079025-35079025 on Build GRCh38
rs199676237 Chr.14: 35078978-35078978 on Build GRCh38
rs951531869 Chr.14: 35079018-35079018 on Build GRCh38
rs746856414 Chr.14: 35079069-35079069 on Build GRCh38
rs992834211 Chr.14: 35079093-35079093 on Build GRCh38
rs778219751 Chr.14: 35079051-35079051 on Build GRCh38
rs760449083 Chr.14: 35078988-35078988 on Build GRCh38
rs964507364 Chr.14: 35078995-35078995 on Build GRCh38
rs566651693 Chr.14: 35079099-35079099 on Build GRCh38
rs756514022 Chr.14: 35079033-35079033 on Build GRCh38
rs774072355 Chr.14: 35078946-35078946 on Build GRCh38
rs190706618 Chr.14: 35079117-35079117 on Build GRCh38
rs536518345 Chr.14: 35079011-35079011 on Build GRCh38
rs765450883 Chr.14: 35079041-35079041 on Build GRCh38
rs770501719 Chr.14: 35079047-35079047 on Build GRCh38
rs371724145 Chr.14: 35079056-35079056 on Build GRCh38
rs771905703 Chr.14: 35078968-35078968 on Build GRCh38
rs753755587 Chr.14: 35078996-35078996 on Build GRCh38
rs766524861 Chr.14: 35079052-35079053 on Build GRCh38
rs541185156 Chr.14: 35079118-35079118 on Build GRCh38
rs762116272 Chr.14: 35079030-35079030 on Build GRCh38
rs755101625 Chr.14: 35079024-35079024 on Build GRCh38

View Full Product Details